Skip to Content
Merck
All Photos(1)

Documents

M3324

Sigma-Aldrich

M100907

≥98% (HPLC)

Synonym(s):

(R)-(+)-α-(2,3-dimethoxyphenyl)-1-[2-(4-fluorophenyl)ethyl]-4-pipidinemethanol, MDL100907, Volinanserin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28FNO3
CAS Number:
Molecular Weight:
373.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

COc1cccc([C@H](O)C2CCN(CC2)CCc3ccc(F)cc3)c1OC

InChI

1S/C22H28FNO3/c1-26-20-5-3-4-19(22(20)27-2)21(25)17-11-14-24(15-12-17)13-10-16-6-8-18(23)9-7-16/h3-9,17,21,25H,10-15H2,1-2H3/t21-/m1/s1

InChI key

HXTGXYRHXAGCFP-OAQYLSRUSA-N

Gene Information

human ... HTR2A(3356)

General description

M100907 blocks the serotonin 2A (5-HT2A) receptors and imposes antidepressant-like effects. It may be used for treating dopaminergic anomalies.

Biochem/physiol Actions

M100907 is a selective 5-HT2A Antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela J Rylands et al.
Biological psychiatry, 72(12), 1004-1011 (2012-07-28)
Impulsive aggression (IA) in adults is associated with brain serotonin (5-HT) system abnormalities and is more common following childhood adversity. Within aggressive behavior, IA and callous unemotional (CU) traits are core components of differentiable factors with opposing 5-HT abnormalities. We
Ashley Furr et al.
The international journal of neuropsychopharmacology, 15(9), 1295-1305 (2011-10-20)
Chronic stress is a risk factor for depression, and chronic stress can induce cognitive impairments associated with prefrontal cortical dysfunction, which are also major components of depression. We have previously shown that 5 wk chronic intermittent cold (CIC) stress induced
Kerry A Waters et al.
Behavioural brain research, 228(1), 211-218 (2011-12-23)
The 5-hydroxytryptamine7 (5-HT7) receptor is a G-protein coupled receptor for serotonin that has been implicated in the pathophysiology of psychiatric and neurological disorders including anxiety, depression and schizophrenia. A number of studies have attempted to evaluate the potential role of
Lara A Pockros et al.
Psychopharmacology, 213(2-3), 307-320 (2010-11-17)
The action of serotonin (5-HT) at the 5-HT(2A) receptor subtype is thought to be involved in cocaine-seeking behavior that is motivated by exposure to drug-associated cues and drug priming. 5-HT(2A) receptors are densely clustered in the ventromedial prefrontal cortex (vmPFC)
Mona E Hervig et al.
Psychology & neuroscience, 13(3), 438-458 (2021-02-23)
Adapting behavior to a dynamic environment requires both steadiness when the environment is stable and behavioral flexibility in response to changes. Much evidence suggests that cognitive flexibility, which can be operationalized in reversal learning tasks, is mediated by cortico-striatal circuitries

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service