Skip to Content
Merck
All Photos(1)

Documents

M2910

Sigma-Aldrich

[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11

≥97% (HPLC)

Synonym(s):

GHRP-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H56N12O6
CAS Number:
Molecular Weight:
873.01
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@@H](N)Cc3cnc[nH]3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@H](Cc6ccccc6)C(=O)N[C@@H](CCCCN)C(N)=O

InChI

1S/C46H56N12O6/c1-27(54-44(62)39(20-29-23-51-35-15-7-5-13-32(29)35)57-43(61)34(48)22-31-25-50-26-53-31)42(60)56-40(21-30-24-52-36-16-8-6-14-33(30)36)46(64)58-38(19-28-11-3-2-4-12-28)45(63)55-37(41(49)59)17-9-10-18-47/h2-8,11-16,23-27,34,37-40,51-52H,9-10,17-22,47-48H2,1H3,(H2,49,59)(H,50,53)(H,54,62)(H,55,63)(H,56,60)(H,57,61)(H,58,64)/t27-,34-,37-,38+,39+,40-/m0/s1

InChI key

WZHKXNSOCOQYQX-FUAFALNISA-N

Gene Information

human ... POMC(5443)
rat ... Ghsr(84022)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Trp-Ala-Trp-Phe-Lys-NH2

Biochem/physiol Actions

Selective inhibitor of α-MSH activity in frog skin bioassay; identical to (His1,Lys8)-GH which selectively releases growth hormone in vitro and in vivo; somatostatin antagonist.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessia Indrieri et al.
EMBO molecular medicine, 5(2), 280-293 (2012-12-15)
Mitochondrial-dependent (intrinsic) programmed cell death (PCD) is an essential homoeostatic mechanism that selects bioenergetically proficient cells suitable for tissue/organ development. However, the link between mitochondrial dysfunction, intrinsic apoptosis and developmental anomalies has not been demonstrated to date. Now we provide
Diana García Del Barco-Herrera et al.
Restorative neurology and neuroscience, 31(2), 213-223 (2013-01-15)
Stroke is the second cause of mortality worldwide, with a high incidence of disability in survivors. Promising candidate drugs have failed in stroke trials. Combined therapies are attractive strategies that simultaneously target different points of stroke pathophysiology. The aim of
Fernando Guerrero et al.
PloS one, 7(11), e50470-e50470 (2012-11-28)
The ethylene-forming enzyme (EFE) from Pseudomonas syringae catalyzes the synthesis of ethylene which can be easily detected in the headspace of closed cultures. A synthetic codon-optimized gene encoding N-terminal His-tagged EFE (EFEh) was expressed in Synechocystis sp. PCC 6803 (Synechocystis)
Magdalena Arasimowicz-Jelonek et al.
Plant physiology and biochemistry : PPB, 63, 177-184 (2012-12-26)
Homocysteine (Hcy) is a naturally occurring intermediate metabolite formed during methionine metabolism. It has been well documented that its excess can be extremely toxic to mammalian, yeast and bacterial cells. In spite of the metabolic value of Hcy known for
Marylou Machingura et al.
Plant physiology and biochemistry : PPB, 63, 159-169 (2012-12-25)
The β-cyanoalanine pathway is primarily responsible for detoxification of excess cyanide produced by plants. Recent evidence suggests that cyanide detoxification via this pathway may be involved in the response and tolerance to water deficit in plants. The aim of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service