Skip to Content
Merck
All Photos(3)

Documents

M2140

Sigma-Aldrich

1-Decanoyl-rac-glycerol

≥99%

Synonym(s):

Monocaprin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26O4
CAS Number:
Molecular Weight:
246.34
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder

functional group

ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(OCC(O)CO)CCCCCCCCC

InChI

1S/C13H26O4/c1-2-3-4-5-6-7-8-9-13(16)17-11-12(15)10-14/h12,14-15H,2-11H2,1H3

InChI key

LKUNXBRZDFMZOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Novel LC/MS/MS and High-Throughput Mass Spectrometric Assays for Monoacylglycerol Acyltransferase Inhibitors.: This research presents advanced assays for screening inhibitors of monoacylglycerol acyltransferase, with implications for metabolic disease treatment. 1-Decanoyl-rac-glycerol is a key compound in the study (Qi J et al., 2017).

  • Optimized reverse micelle surfactant system for high-resolution NMR spectroscopy of encapsulated proteins and nucleic acids dissolved in low viscosity fluids.: The paper describes a novel surfactant system using 1-Decanoyl-rac-glycerol, which improves the NMR spectroscopy of proteins and nucleic acids, offering better resolution and reduced viscosity (Dodevski I et al., 2014).

Biochem/physiol Actions

Monocaprin (glycerol monocaprate) is used in the development of antimicrobial disinfectants active against enveloped viruses, and certain yeast and bacteria such as candida albicans and campylobacter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thórunn Osk Thorgeirsdóttir et al.
Acta odontologica Scandinavica, 64(1), 21-26 (2006-01-24)
Monocaprin is a 1-monoglyceride of capric acid that has antimicrobial activity against enveloped viruses, certain bacteria, and the yeast Candida albicans. Solutions containing monocaprin were formulated and tested in vitro against a number of micro-organisms, including species found in the
T Kristmundsdóttir et al.
Drug development and industrial pharmacy, 29(2), 121-129 (2003-03-22)
Hydrogel formulations containing the monoglyceride monocaprin have shown potent microbicidal activity against several sexually transmitted viruses and bacteria. It is recommended that formulations for preventing infection in the vagina have a low pH as the HIV virus is inactivated at
Sandra L Blair et al.
Journal of the American Society for Mass Spectrometry, 29(4), 635-639 (2018-02-07)
In this communication, we report on the real-time analysis of organic aerosol particles by Vacuum-assisted Plasma Ionization-Mass Spectrometry (Aero-VaPI-MS) using a home-built VaPI ion source coupled to a Synapt G2-S HDMS ion mobility-mass spectrometry (IM-MS) system. Standards of organic molecules
Halldor Thormar et al.
Applied and environmental microbiology, 72(1), 522-526 (2006-01-05)
Of 11 fatty acids and monoglycerides tested against Campylobacter jejuni, the 1-monoglyceride of capric acid (monocaprin) was the most active in killing the bacterium. Various monocaprin-in-water emulsions were prepared which were stable after storage at room temperature for many months
Victoria Dolange et al.
Antibiotics (Basel, Switzerland), 7(3) (2018-07-14)
Antibiotic-resistant gonorrhea is now a reality, as well as the consequences of untreatable infections. Gonococcal eye infections result in blindness if not properly treated; they accounted for the vast majority of infections in children in homes for the blind in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service