Skip to Content
Merck
All Photos(3)

Key Documents

M1640

Sigma-Aldrich

DL-α-Palmitin

≥99%

Synonym(s):

MG(16:0/0:0/0:0)[rac], α-Monopalmitin, 1-Monohexadecanoyl-rac-glycerol, rac-1-Palmitoylglycerol, rac-Glycerol 1-palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OCO(CH2)14CH3
CAS Number:
Molecular Weight:
330.50
Beilstein:
1728235
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant
synthetic (organic)

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(O)CO

InChI

1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,20-21H,2-17H2,1H3

InChI key

QHZLMUACJMDIAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Monopalmitin is a glycerol esterified fatty acid, monoacylglycerol, found in lipid structures such as ceramides. 1-Palmitoylglycerol may be used as a substrate for the identification and differentiation of enzymes that hydrolyze or transfer monoacylglycerols such as neuropathy target esterase(s) and monoacylglycerol acyltransferase(s) (MAG).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica A Conlan et al.
PloS one, 12(11), e0188568-e0188568 (2017-11-29)
Heterotrophic feeding in newly-settled coral planulae can potentially improve survivorship and accelerate early development in some species; however, an optimal diet to facilitate this does not currently exist. This study evaluated the efficacy of three heterotrophic feeding regimes (enriched rotifers
Jessica A Conlan et al.
PeerJ, 6, e4239-e4239 (2018-02-07)
Scleractinian corals are colonial organisms comprising multiple physiologically integrated polyps and branches. Colonialism in corals is highly beneficial, and allows a single colony to undergo several life processes at once through physiological integration and compartmentalised functioning. Elucidating differences in the
Jessica A Conlan et al.
PeerJ, 5, e3645-e3645 (2017-08-09)
Lipids are involved in a host of biochemical and physiological processes in corals. Therefore, changes in lipid composition reflect changes in the ecology, nutrition, and health of corals. As such, accurate lipid extraction, quantification, and identification is critical to obtain
Yin-Ku Lin et al.
International journal of nanomedicine, 5, 117-128 (2010-03-24)
The combination of calcipotriol with methotrexate can strengthen the topical therapy for psoriasis. The aim of the present study was to evaluate the potential of nanostructured lipid carriers (NLCs) loaded with lipophilic calcipotriol and hydrophilic methotrexate as topical therapy. NLCs
J M Brewer et al.
Immunology, 75(4), 570-575 (1992-04-01)
The ability of non-ionic surfactant vesicles (NISV) to enhance antibody production against bovine serum albumin (BSA) was compared with Freund's complete adjuvant (FCA), in the BALB/c mouse. Two subcutaneous inoculations with NISV entrapped BSA induced antibody levels comparable to, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service