Skip to Content
Merck
All Photos(1)

Key Documents

M1317

Sigma-Aldrich

MES solution

BioPerformance Certified, suitable for, 1 M, suitable for cell culture

Synonym(s):

2-(N-Morpholino) ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified
for molecular biology

Quality Level

sterility

sterile; 0.2 μm filtered

form

solution

concentration

1 M

technique(s)

cell culture | mammalian: suitable

useful pH range

5.5-6.7

cation traces

Fe: <5 ppm

suitability

suitable for

storage temp.

room temp

SMILES string

[S](=O)(=O)(O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

Application

MES solution has been used to activate microspheres. It has also been used as a component of activating solution to activate carboxyl groups of poly lactic-co-glycolic acid (PLGA).

Other Notes

May form precipitate at low temperatures, heat to redissolve (40 °C for 30 minutes).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of quantitative suspension array assays for six immunoglobulin isotypes and subclasses to multiple Plasmodium falciparum antigens
Vidal M, et al.
Journal of Immunological Methods, 455, 41-54 (2018)
Quantification of Confocal images using LabVIEW for tissue engineering applications
Sfakis L, et al.
Tissue Engineering: Part C, Methods, 22(11), 1028-1037 (2016)
Ariel Rodriguez-Frandsen et al.
Journal of virology, 94(4) (2019-11-30)
Influenza A virus (IAV) is a human respiratory pathogen that causes yearly global epidemics, as well as sporadic pandemics due to human adaptation of pathogenic strains. Efficient replication of IAV in different species is, in part, dictated by its ability
Saurabh Raj et al.
Biomedical optics express, 3(4), 753-763 (2012-05-11)
Two microparticles were biochemically attached to a red blood cell at diametrically opposite parts and held by optical traps allowing to impose deformations. The cell deformation was monitored from the microscopy images. Raman spectra of the cell under tunable deformations
Zahraa I Foraida et al.
Acta biomaterialia, 62, 116-127 (2017-08-13)
Development of electrospun nanofibers that mimic the structural, mechanical and biochemical properties of natural extracellular matrices (ECMs) is a promising approach for tissue regeneration. Electrospun fibers of synthetic polymers partially mimic the topography of the ECM, however, their high stiffness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service