M1126
L-Methionine sulfoxide
≥98% (TLC)
Synonym(s):
L-2-Amino-4-(methylsulfinyl)butanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H11NO3S
CAS Number:
Molecular Weight:
165.21
Beilstein:
4129939
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
L-Methionine sulfoxide,
Assay
≥98% (TLC)
form
powder
color
white to off-white
mp
255 °C
application(s)
detection
peptide synthesis
SMILES string
CS(=O)CC[C@H](N)C(O)=O
InChI
1S/C5H11NO3S/c1-10(9)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-,10?/m0/s1
InChI key
QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Gene Information
rat ... Ggt1(116568)
Looking for similar products? Visit Product Comparison Guide
Application
- The DmsABC S-oxide reductase is an essential component of a novel, hypochlorite-inducible system of extracellular stress defense in Haemophilus influenzae: This study illustrates the enzymatic action of L-Methionine sulfoxide reductase in bacterial defense systems, underscoring its biotechnological applications in developing antimicrobial strategies (Nasreen et al., 2024).
Biochem/physiol Actions
L-Methionine sulfoxide, a structural analog of glutamate, may be used as a substrate to study the specificity and kinetics of various methionine sulfoxide reductase(s).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Martina I Boxler et al.
Journal of proteome research, 16(9), 3310-3320 (2017-07-20)
3,4-Methylenedioxymethamphetamine (MDMA; "ecstasy") is widely consumed recreationally. Little is known about its effects on the human metabolome. Mapping biochemical changes after drug exposure can complement traditional approaches by revealing potential biomarkers of organ toxicity or discovering new metabolomic features in
Gabriel Javitt et al.
Antioxidants & redox signaling, 33(10), 665-678 (2020-06-11)
Aims: The post-translational oxidation of methionine to methionine sulfoxide (MetSO) is a reversible process, enabling the repair of oxidative damage to proteins and the use of sulfoxidation as a regulatory switch. MetSO reductases catalyze the stereospecific reduction of MetSO. One
Jung Min Kim et al.
Scientific reports, 7(1), 11642-11642 (2017-09-16)
Despite significant advances on fluorescent labeling of target proteins to study their structural dynamics and function, there has been need for labeling with high quantum yield ensuring high sensitivity and selectivity without sacrificing the biological function of the protein. Here
Benjamin P Barré et al.
Genome research, 30(5), 697-710 (2020-04-12)
Aging varies among individuals due to both genetics and environment, but the underlying molecular mechanisms remain largely unknown. Using a highly recombined Saccharomyces cerevisiae population, we found 30 distinct quantitative trait loci (QTLs) that control chronological life span (CLS) in
Sofia Lourenço Dos Santos et al.
Antioxidants (Basel, Switzerland), 7(12) (2018-12-14)
Cysteine and methionine residues are the amino acids most sensitive to oxidation by reactive oxygen species. However, in contrast to other amino acids, certain cysteine and methionine oxidation products can be reduced within proteins by dedicated enzymatic repair systems. Oxidation
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service