Skip to Content
Merck
All Photos(1)

Key Documents

M0660

Sigma-Aldrich

Mycoplasma Agar

powder, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

form

powder

solubility

H2O: 35.5 g/L

application(s)

microbiology

General description

Boil to dissolve the agar and adjust pH to 7.8±0.2. Autoclave at 121°C, 15 psi for 15 minutes. Store prepared medium at 0-5°C.
The components contained in this basal medium do not exhibit inhibitory or toxic effects when used to culture mycoplasma. A special mineral supplement has been added to improve the growth and colony characteristics of the organism.

Application

Recommended for the cultivation of mycoplasma

Components

Powder contains (g/L): 10.0 bacterial peptone, 10.0 LAB-LEMCO powder, 5.0 sodium chloride, 0.5 mineral supplement, 10.0 agar.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service