M0535
Mycoplasma Broth
powder, suitable for microbiology
Synonym(s):
Mycoplasma media
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106200
NACRES:
NA.74
Recommended Products
form
powder
technique(s)
microbiological culture: suitable
solubility
H2O: 25.5 g/L (Autoclave at 121°C, 15 psi for 15 minutes. Store prepared medium at 0-5°C.)
application(s)
microbiology
Related Categories
General description
Mycoplasma Broth is a basal medium which, after enrichment with yeast extract, horse serum, and antibiotics, will support the growth of Mycoplasma sp. The components contained in this basal medium do not exhibit inhibitory or toxic effects when used to culture mycoplasma. A special mineral supplement has been added to improve the growth and colony characteristics of the organism.
Application
Mycoplasma Broth has been used for isolation and cultivation of mycoplasma. It may also be used to grow Porphyromonas gingivalis under anerobic conditions.
Components
Powder contains (g/L): 10.0 bacterial peptone, 10.0 LAB-LEMCO powder, 5.0 sodium chloride, 0.5 mineral supplement.
Preparation Note
Basal Medium:
1. Suspend 25.5 g of powdered medium in 1L of distilled water. Stir until the powder dissolves.
2. Adjust the pH of the solution to 7.8 ± 0.2.
3. Autoclave at 121 °C and 15 psi for 15 minutes to sterilize. Cool to 50 °C and add sterile supplements.
1. Suspend 25.5 g of powdered medium in 1L of distilled water. Stir until the powder dissolves.
2. Adjust the pH of the solution to 7.8 ± 0.2.
3. Autoclave at 121 °C and 15 psi for 15 minutes to sterilize. Cool to 50 °C and add sterile supplements.
Storage and Stability
Store dry powdered medium at room temperature and prepare medium at 2-8 °C.
Other Notes
The product is intended for laboratory use only.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amir H Abd-Elrahman et al.
Tropical animal health and production, 52(3), 1179-1186 (2019-11-11)
Contagious caprine pleuropneumonia (CCPP) is one of the most fatal and contagious diseases of goats. To date, the occurrence of CCPP in Egypt has not been reported. During the period from 2017 to 2018, 200 goats and 400 sheep from
Philip Stashenko et al.
The American journal of pathology, 170(1), 203-213 (2007-01-04)
Bacterial infections of the dental pulp result in soft tissue and alveolar bone destruction. It has been suggested that Th1 responses promote disease, whereas Th2 responses are protective. However, other studies have challenged this notion. To address this question, bone
Davor Ivankovic et al.
Autophagy, 16(3), 391-407 (2019-05-31)
Adaptor protein (AP) complexes mediate key sorting decisions in the cell through selective incorporation of transmembrane proteins into vesicles. Little is known of the roles of AP-4, despite its loss of function leading to a severe early onset neurological disorder
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service