Skip to Content
Merck
All Photos(4)

Key Documents

L9781

Sigma-Aldrich

Lithium dodecyl sulfate

BioReagent, for molecular biology, suitable for electrophoresis

Synonym(s):

Dodecyl lithium sulfate, Dodecyl sulfate lithium salt, Lauryl sulfate lithium salt, Lithium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Li
CAS Number:
Molecular Weight:
272.33
Beilstein:
5186718
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.31

grade

Molecular Biology
for molecular biology

description

anionic

product line

BioReagent

Assay

≥98.5% (GC)

mol wt

272.33 g/mol

CMC

7-10 mM (20-25°C)

transition temp

cloud point >100 °C

absorption

≤0.05 at 260 nm (3% solution)
≤0.05 at 280 nm (3% solution)

suitability

suitable for electrophoresis

foreign activity

DNase and RNase, none detected

SMILES string

[Li+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Li/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

YFVGRULMIQXYNE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent that may be used in place of SDS for electrophoresis in cold conditions.
Lithium dodecyl sulfate has been used as a component of lysis buffer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

System-wide identification of RNA-binding proteins by interactome capture
Castello A, et al.
Nature Protocols, 8(3), 491-491 (2013)
Ultra-high-throughput sequencing of the immune receptor repertoire from millions of lymphocytes
McDaniel J R, et al.
Nature Protocols, 11(3), 429-429 (2016)
Eric J Montemayor et al.
Nucleic acids research, 42(16), 10845-10855 (2014-08-16)
The enzymatic processing of cellular RNA molecules requires selective recognition of unique chemical and topological features. The unusual 2',5'-phosphodiester linkages in RNA lariats produced by the spliceosome must be hydrolyzed by the intron debranching enzyme (Dbr1) before they can be
Michael Backlund et al.
Nucleic acids research, 48(9), 4725-4740 (2020-04-22)
Cellular stress causes multifaceted reactions to trigger adaptive responses to environmental cues at all levels of the gene expression pathway. RNA-binding proteins (RBP) are key contributors to stress-induced regulation of RNA fate and function. Here, we uncover the plasticity of
F Bruna et al.
Journal of hazardous materials, 225-226, 74-80 (2012-05-23)
Polycyclic aromatic hydrocarbons (PAHs) are considered as priority pollutants because of their high risk to human health. In this paper, we addressed the issue of using hydrotalcite-based nanocomposites as adsorbents of six low molecular weight PAHs (acenaphthene, fluorene, phenanthrene, anthracene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service