Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium dodecyl sulfate
Synonym(s):
Dodecyl lithium sulfate, Dodecyl sulfate lithium salt, Lauryl sulfate lithium salt, Lithium lauryl sulfate
Linear Formula:
CH
3
(CH
2
)
11
OSO
3
Li
CAS Number:
2044-56-6
Molecular Weight:
272.33
EC Number:
218-058-2
Products
Product Category
detergents (3)
Brand
Sigma-Aldrich (3)
Feature
Formula Weight
Product Line
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
L9781
Lithium dodecyl sulfate,
BioReagent, for molecular biology, suitable for electrophoresis
Expand
View Pricing
L4632
Lithium dodecyl sulfate,
≥98.5% (GC)
Expand
View Pricing
L5901
Lithium dodecyl sulfate,
BioXtra, ≥98.5% (GC)
Expand
View Pricing