L7634
α-Lactose-Agarose
saline suspension
Synonym(s):
alpha-lactose resin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine milk
form
saline suspension
technique(s)
affinity chromatography: suitable
matrix
6% beaded agarose
matrix activation
divinyl sulfone
matrix attachment
hydroxyl
matrix spacer
5 atoms
capacity
6-25 mg/mL binding capacity (lectin from Arachis hypogaea)
suitability
suitable for chromatography
storage temp.
2-8°C
Application
α-Lactose-agarose has been used:
- in protein chromatography to purify galectins from lysed bacteria cells
- to capture ricin based on lectin-carbohydrate affinity
- to study microneme proteins and host-parasite cell surface receptors
α-lLactose-agarose is used in protein chromatography, affinity chromatography and carbohydrate matrices. α--Lactose-agarose has been used to study microneme proteins and host-parasite cell surface receptors.
Physical form
Suspension in 0.15 M NaCl, 0.01 M sodium phosphate, pH 6.8, containing 0.02% sodium azide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Christophe Dussouy et al.
Chemical biology & drug design, 96(4), 1123-1133 (2020-03-29)
Galectins play key roles in numerous biological processes. Their mode of action depends on their localization which can be extracellular, cytoplasmic, or nuclear and is partly mediated through interactions with β-galactose containing glycans. Galectins have emerged as novel therapeutic targets
C Allison Gray et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(21), 7982-7987 (2004-05-19)
Secretions of the uterus support survival and growth of the conceptus (embryo/fetus and associated membranes) during pregnancy. Galectin-15, also known as OVGAL11 and a previously uncharacterized member of the galectin family of secreted beta-galactoside lectins containing a conserved carbohydrate recognition
E V Lourenço et al.
Glycobiology, 11(7), 541-547 (2001-07-12)
Host cell invasion by Toxoplasma gondii is a multistep process with one of the first steps being the apical release of micronemal proteins that interact with host receptors. We demonstrate here that micronemal protein 1 (MIC1) is a lactose-binding lectin.
Nadine Keller et al.
Infection and immunity, 72(8), 4791-4800 (2004-07-24)
Microneme proteins have been shown to play an important role in the early phase of host cell adhesion, by mediating the contact between the parasite and host cell surface receptors. In this study we have identified and characterized a lectin-like
Na Yang et al.
Journal of molecular biology, 387(3), 694-705 (2009-04-14)
Lectin AAL (Agrocybe aegerita lectin) from the edible mushroom A. aegerita is an antitumor protein that exerts its tumor-suppressing function via apoptosis-inducing activity in cancer cells. The crystal structures of ligand-free AAL and its complex with lactose have been determined.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service