Skip to Content
Merck
All Photos(1)

Key Documents

V3700

Sigma-Aldrich

Divinyl sulfone

contains hydroquinone as inhibitor, ≥96%

Synonym(s):

Vinyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CH)2SO2
CAS Number:
Molecular Weight:
118.15
Beilstein:
1071329
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥96%

contains

hydroquinone as inhibitor

refractive index

n20/D 1.476 (lit.)

bp

234 °C (lit.)

mp

−26 °C (lit.)

density

1.177 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CS(=O)(=O)C=C

InChI

1S/C4H6O2S/c1-3-7(5,6)4-2/h3-4H,1-2H2

InChI key

AFOSIXZFDONLBT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Divinyl sulfone is a highly reactive chemical compound due to the presence of two vinyl groups, which make it an effective cross-linking agent and bifunctional reactive compound. It is used in the production of hydrogels, resins, and other polymeric materials. The resulting cross-linked polymers exhibit enhanced mechanical strength, stability, and insolubility in solvents.

Application

Divinyl sulfone can be used as:
  • A cross-linking agent to synthesize divinyl sulfone-crosslinked hyaluronic acid hydrogels for specific biomedical applications, such as tissue engineering or drug delivery.
  • A cross-linking agent to develop the conducting polymer film with MXene layers. This crosslinking can enhance the mechanical properties and stability of the composite film.
Hyaluronic acid (HA) crosslinked with DVS forms hydrogels.
DVS and its mono and di-substituted derivatives are useful starting materials in the preparation of thiomorpholine 1,1-dioxides and other synthetically important macro- and the heterocycles.
DVS may be used to shrink proofing cotton by crosslinking it with cellulose.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Käppler et al.
Biotechnology and bioengineering, 102(2), 535-545 (2008-08-30)
A new approach for in situ product removal from bioreactors is presented in which high-gradient magnetic separation is used. This separation process was used for the adsorptive removal of proteases secreted by Bacillus licheniformis. Small, non-porous bacitracin linked magnetic adsorbents
Stereocontrolled creation of all-carbon quaternary stereocenters by organocatalytic conjugate addition of oxindoles to vinyl sulfone.
Qiang Zhu et al.
Angewandte Chemie (International ed. in English), 49(42), 7753-7756 (2010-09-08)
Adrien Quintard et al.
Chemical communications (Cambridge, England), 47(25), 7212-7214 (2011-05-24)
An unprecedented intermolecular iminium/enamine Michael addition on enals has been developed by taking advantage of the high reactivity of vinyl sulfones. This powerful organocascade allows for the rapid construction of attractive synthons in high enantioselectivities (typically 99% ee).
Yan Li et al.
Biomaterials, 33(27), 6533-6541 (2012-06-19)
Effective delivery of DNA to direct cell behavior in a well defined three dimensional scaffold offers a superior approach in tissue engineering. In this study, we synthesized biodegradable nanostructured hydrogels with tunable physical properties for cell and gene delivery. The
Elizabeth Dunny et al.
Journal of medicinal chemistry, 56(17), 6638-6650 (2013-08-21)
A series of vinyl sulfone-containing peptidomimetics were rationally designed, synthesized, and evaluated against Trypanosoma brucei brucei . These electrophilic compounds are likely to exert their antitrypanosomal activity via inhibition of trypanosomal cysteine proteases, TbCatB and rhodesain, through alkylation of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service