Skip to Content
Merck
All Photos(1)

Key Documents

L3916

Sigma-Aldrich

L-Lactic Dehydrogenase from bovine heart

1000 units/mL

Synonym(s):

Lactate, (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

solution

Quality Level

specific activity

1000 units/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

LDH (lactic dehydrogenase) is a glycolytic enzyme. It is found in skeletal muscle, heart, liver, kidneys, brain, lungs and red blood cells. It comprises of five isoenzyme forms. LDH possess a tetrameric structure.

Application

L-Lactic Dehydrogenase from bovine heart has been used:
  • as a reference standard to measure the LDH (lactic dehydrogenase)
  • to quench protein-bound NADH in cells
  • to measure the lactate released by the cells in suspension

For use in enzymatic determination of lactate or pyruvate.

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.
LDH (lactic dehydrogenase) helps in the reversible conversion of lactate to pyruvate.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Suspension in 2.2 M ammonium sulfate

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circulating biomarkers in malignant melanoma
Advances in Clinical Chemistry, 69, 47-89 (2015)
Culture Expansion in Low-Glucose Conditions Preserves Chondrocyte Differentiation and Enhances Their Subsequent Capacity to Form Cartilage Tissue in Three-Dimensional Culture
HeywoodHannah K, et al.
BioResearch Open Access, 2484-2494 (2014)
NADH fluorescence lifetime is an endogenous reporter of alpha-synuclein aggregation in live cells
Plotegher N, et al.
Faseb Journal, 29(6), 2484-2494 (2015)
Mitochondrial angiotensin receptors in dopaminergic neurons. Role in cell protection and aging-related vulnerability to neurodegeneration
Valenzuela R, et al.
Cell Death & Disease, 7(10), e2427-e2427 (2016)
Cardiac, Vascular, and Skeletal Muscle Systems
Haschek and Rousseaux's Handbook of Toxicologic Pathology (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service