Skip to Content
Merck
All Photos(1)

Key Documents

L2625

Sigma-Aldrich

L-Lactic Dehydrogenase from bovine heart

Type III, ammonium sulfate suspension, ≥500 units/mg protein

Synonym(s):

Lactate, (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type III

Quality Level

form

ammonium sulfate suspension

specific activity

≥500 units/mg protein

UniProt accession no.

foreign activity

pyruvate kinase ≤0.03%

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Research area: Cell signaling

Lactate dehydrogenase (LDH) is a cytoplasmic enzyme that belongs to the oxidoreductase’s family. It is ubiquitously expressed in body tissues but at high levels in muscle, liver, and kidney. LDH exists as tetramer and comprises of two subunits (H and M). The LDH of eukaryotes undergo active-site loop gating for their catalytic functionality. L-lactate, which is the major enantiomeric form of lactate in vertebrates, is produced by the L-isomers of LDH enzyme encoded by the LDHC gene.

Application

L-Lactic Dehydrogenase from bovine heart has been used as a standard in cytotoxicity assay. It has also been used in glutamic pyruvic transaminase (GPT) assay.

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.
Lactate dehydrogenase (LDH) is a crucial enzyme that plays a vital role in the anaerobic metabolic pathway. It facilitates the conversion of lactate to pyruvate and NAD⁺ to NADH, and vice versa, within cells. This enzymatic reaction plays a crucial role in cellular energy production, particularly in environments with low oxygen levels or hypoxic conditions. LDH activity is also significant in tumor glycolysis and the Warburg effect. LDH is released into the bloodstream when the cells are damaged or undergo death. Therefore, LDH can be used as a potential marker of cell injury and death. High lactate dehydrogenase activity is associated with various medical conditions, such as liver disease, anemia, heart attack, bone fractures, muscle trauma, and certain types of cancer.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Crystalline suspension in 2.1 M (NH4)2SO4 solution, pH 6.0

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interferon beta-1b inhibits gelatinase secretion and in vitro migration of human T cells: a possible mechanism for treatment efficacy in multiple sclerosis
Leppert D, et al.
Annals of Neurology, 40(6), 846-852 (1996)
Johannes Yayo et al.
Applied and environmental microbiology, 89(1), e0175322-e0175322 (2023-01-11)
Clostridium thermocellum is a cellulolytic thermophile that is considered for the consolidated bioprocessing of lignocellulose to ethanol. Improvements in ethanol yield are required for industrial implementation, but the incompletely understood causes of amino acid secretion impede progress. In this study
Kyoung-Han Kim et al.
The Journal of pharmacology and experimental therapeutics, 324(1), 160-169 (2007-10-12)
Doxorubicin (DOX) is an effective antineoplastic agent whose use has been limited by its cardiotoxic side effects. Recent studies have established that erythropoietin (EPO), a cytokine essential for red blood cell production, protects against ischemic injury in the heart and
Biochemistry, lactate dehydrogenase
Farhana A and Lappin SL
StatPearls [Internet] (2020)
Adaptation of rats to diets containing different levels of protein: effects on food intake, plasma and brain amino acid concentrations and brain neurotransmitter metabolism
Peters JC and Harper AE
The Journal of Nutrition, 115(3), 382-398 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service