Skip to Content
Merck
All Photos(1)

Documents

L2261

Sigma-Aldrich

Lanatoside C

≥97% (TLC)

Synonym(s):

Digilanide C, Digilanogen C, Isolanide, Lanatigen C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H76O20
CAS Number:
Molecular Weight:
985.12
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

C[C@H]1O[C@H](C[C@H](O)[C@@H]1O[C@H]2C[C@H](O)[C@H](O[C@H]3C[C@H](OC(C)=O)[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](C)O3)[C@@H](C)O2)O[C@H]5CC[C@@]6(C)[C@H](CC[C@@H]7[C@@H]6C[C@@H](O)[C@]8(C)[C@H](CC[C@]78O)C9=CC(=O)OC9)C5

InChI

1S/C49H76O20/c1-21-43(67-38-17-32(53)44(22(2)62-38)68-39-18-33(64-24(4)51)45(23(3)63-39)69-46-42(58)41(57)40(56)34(19-50)66-46)31(52)16-37(61-21)65-27-9-11-47(5)26(14-27)7-8-29-30(47)15-35(54)48(6)28(10-12-49(29,48)59)25-13-36(55)60-20-25/h13,21-23,26-35,37-46,50,52-54,56-59H,7-12,14-20H2,1-6H3/t21-,22-,23-,26-,27+,28-,29-,30+,31+,32+,33+,34-,35-,37+,38+,39+,40-,41+,42-,43-,44-,45-,46+,47+,48+,49+/m1/s1

InChI key

JAYAGJDXJIDEKI-PTGWOZRBSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Cardiac glycoside sensitization in hepatocellular carcinoma: Research demonstrated that Lanatoside C enhances the sensitivity of hepatocellular carcinoma cells to TRAIL-induced apoptosis through mechanisms involving reactive oxygen species generation, p38MAPK activation, and modulation of mitochondrial transition and autophagy (Rasheduzzaman et al., 2019).

  • Elimination of undifferentiated human embryonic stem cells: Lanatoside C has been shown to selectively eliminate undifferentiated human embryonic stem cells, indicating its potential use in ensuring the safety of stem cell-derived therapies (Lin et al., 2017).

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Ueda et al.
The Journal of organic chemistry, 77(18), 7850-7857 (2012-08-09)
Acylation of lanatoside C in the presence of organocatalyst 5 gave the C(4'''')-O-acylate in up to 90% regioselectivity (catalyst-controlled regioselectivity). Various functionalized acyl groups can be introduced at the C(4'''')-OH by a mixed anhydride method in the presence of 5
T Imamura et al.
Circulation, 71(1), 11-16 (1985-01-01)
We sought to determine whether digitalis augments cardiopulmonary baroreflex control of forearm vascular resistance in normal young men. Cardiopulmonary baroreceptor input was reduced with lower body negative pressure (LBNP) at 10 and 20 mm Hg, which decreased central venous pressure
Christian E Badr et al.
Neuro-oncology, 13(11), 1213-1224 (2011-07-16)
Human glioblastoma (GBM) cells are notorious for their resistance to apoptosis-inducing therapeutics. We have identified lanatoside C as a sensitizer of GBM cells to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)-induced cell death partly by upregulation of the death receptor 5.
Dhanasekhar Reddy et al.
Biomolecules, 9(12) (2019-12-01)
Cardiac glycosides (CGs) are a diverse family of naturally derived compounds having a steroid and glycone moiety in their structures. CG molecules inhibit the α-subunit of ubiquitous transmembrane protein Na+/K+-ATPase and are clinically approved for the treatment of cardiovascular diseases.
A Takeshita et al.
Japanese circulation journal, 50(7), 671-674 (1986-07-01)
We examined whether digitalis augments cardiopulmonary baroreflex control of forearm vascular resistance in normal young men. Cardiopulmonary baroreceptor input was reduced with lower body negative pressure (LBNP) at 10 and 20 mmHg which decreased central venous pressure (CVP) but did

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service