Skip to Content
Merck
All Photos(1)

Key Documents

S6626

Sigma-Aldrich

Strophanthidin

Synonym(s):

3β,5,14-Trihydroxy-19-oxo-5β,20(22)-cardenolide, Apocymarin, Convallatoxigenin, Corchorin, Corchoside A aglycone, Corchsularin, Cymarigenen, Erysimupicrone, K-Strophanthidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

storage temp.

−20°C

SMILES string

C[C@@]12[C@@](CC[C@@H]2C(CO3)=CC3=O)(O)[C@]4([H])CC[C@]5(O)C[C@@H](O)CC[C@]5(C([H])=O)[C@@]4([H])CC1

InChI

1S/C23H32O6/c1-20-6-3-17-18(4-8-22(27)11-15(25)2-7-21(17,22)13-24)23(20,28)9-5-16(20)14-10-19(26)29-12-14/h10,13,15-18,25,27-28H,2-9,11-12H2,1H3

InChI key

ODJLBQGVINUMMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strophanthidin was used to synthesize acetylstrophanthidin and the effect on neurotransmitter release from dog saphenous vein was studied.3

Biochem/physiol Actions

Strophanthidin is a cardiotonic steroid that elevates the activity of Na+/K+-ATPase in cardiac myocytes.1 It decreases the potassium content and raises the sodium content in rabbit renal cortex slices.2

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark R Fowler et al.
Cardiovascular research, 62(3), 529-537 (2004-05-26)
Recent work has suggested that Na(+)/Ca(2+) exchange (NCX) and L-type Ca(2+) current (I(Ca)) are located predominantly in the t-tubules of cardiac ventricular myocytes, which therefore represent a microdomain for the regulation of intracellular Na(+) (Na(i)) and Ca(2+) (Ca(i)). The aim
Sara Arganda et al.
Nature neuroscience, 10(11), 1467-1473 (2007-10-02)
Pump activity is a homeostatic mechanism that maintains ionic gradients. Here we examined whether the slow reduction in excitability induced by sodium-pump activity that has been seen in many neuronal types is also involved in neuronal coding. We took intracellular
Dirk von Lewinski et al.
European journal of heart failure, 9(11), 1086-1094 (2007-10-25)
Cardiac glycosides are characterized by a narrow therapeutic range with Ca2+-overload and arrhythmias occurring at higher concentrations. Data on cardiac glycosides in isolated failing human myocardium are scarce and the frequency-dependent actions and toxicity of Strophanthidin have not yet been
Beni Shpak et al.
European journal of pharmacology, 553(1-3), 196-204 (2006-11-03)
Previous studies demonstrated that the purified endogenous inhibitor (NCX(IF)) of the cardiac Na(+)/Ca(2+) exchanger (NCX1) has the capacity to modulate cardiac muscle contractility. Here, we tested the effects of purified NCX(IF) on arrhythmias induced by ouabain in the atria and
Dhanasekhar Reddy et al.
Frontiers in oncology, 9, 1469-1469 (2020-02-06)
Lung cancer is the most prevalent in cancer-related deaths, while breast carcinoma is the second most dominant cancer in women, accounting for the most number of deaths worldwide. Cancers are heterogeneous diseases that consist of several subtypes based on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service