Skip to Content
Merck
All Photos(1)

Key Documents

I9135

Sigma-Aldrich

Anti-Human IgG (Fc specific) antibody produced in rabbit

whole antiserum

Synonym(s):

Human Antiserum, Rabbit Anti Human IgG - Anti-Human IgG (Fc specific) antibody produced in rabbit, Rabbit Anti Human Igg, Rabbit Antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:100,000
quantitative precipitin assay: 1.5 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

IgG contains four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, that are associated by inter-chain disulfide bonds. IgG contains four isotypes IgG1, IgG2, IgG3 and IgG4.

Application

Anti-Human IgG (Fc specific) antibody produced in rabbit was used in immunoblotting and immunocytochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

IgG activates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a pivotal role in antibody-dependent cell-mediated cytotoxicity (ADCC) and phagocytosis. It might be linked to allergy and has the longest serum half life.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Preparation Note

treated to remove lipoproteins

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy William et al.
Molecular psychiatry (2021-06-26)
Suicide is a significant public health concern with complex etiology. Although the genetic component of suicide is well established, the scope of gene networks and biological mechanisms underlying suicide has yet to be defined. Previously, we reported genome-wide evidence that
John D Bartlett et al.
Cells, tissues, organs, 194(2-4), 222-226 (2011-04-29)
Dental enamel development occurs in stages as observed by the changing morphology of the ameloblasts that are responsible for enamel formation. During the secretory stage of development, proteins including MMP20 are secreted into the enamel matrix. MMP20 is required for
Puja Gupta et al.
BMC biotechnology, 10, 72-72 (2010-10-12)
EGF receptor variant III (EGFRvIII) is the most common variant of the EGF receptor observed in human tumors. It results from the in frame deletion of exons 2-7 and the generation of a novel glycine residue at the junction of
A revised nomenclature for allergy: an EAACI position statement from the EAACI nomenclature task force
Johansson SGO, et al.
Allergy, 56(9), 813-824 (2001)
Franco Klingberg et al.
The Journal of cell biology, 207(2), 283-297 (2014-10-22)
Integrin-mediated force application induces a conformational change in latent TGF-β1 that leads to the release of the active form of the growth factor from the extracellular matrix (ECM). Mechanical activation of TGF-β1 is currently understood as an acute process that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service