Skip to Content
Merck
All Photos(1)

Key Documents

I7784

Sigma-Aldrich

Anti-Interleukin-2 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, mouse

technique(s)

neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Il2(16183)
rat ... Il2(116562)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Interleukin-2 (IL-2) is a cytokine produced by activated T cells in response to antigen stimulation. IL-2 is important for proliferation of lymphocytes, natural killer cells and macrophages and in the differentiation of CD4+ cells into Th1 and Th2 effector cells. The effects of IL-2 are mediated by the signalling activated by its binding to IL-2 receptor (IL-2 R). IL-2 activates key signalling pathways such as MAPK, JAK/STAT as well as PI3K/Akt. Activation of these pathways modulates production of inflammatory cytokines, survival and progression through cell cycle
Monoclonal Anti-Interleukin-2 recognizes murine Interleukin-2. It does not react with human IL-2, but will react with rabbit IL-2 at a 100x higher IgG concentration.

Immunogen

purified, E. coli-derived recombinant mouse interleukin-2.

Application

Anti-interleukin-2 antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. The antibody is suitable for murine IL-2 neutralization assays.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew A Williams et al.
Nature, 441(7095), 890-893 (2006-06-17)
Although interleukin-2 (IL-2) was initially characterized as the primary T-cell growth factor following in vitro activation, less is known about its role in shaping T-cell responses to acute infections in vivo. The use of IL-2- or IL-2-receptor-deficient mice is problematic
B H Nelson et al.
Advances in immunology, 70, 1-81 (1998-10-02)
Studies of the biology of the IL-2 receptor have played a major part in establishing several of the fundamental principles that govern our current understanding of immunology. Chief among these is the contribution made by lymphokines to regulation of the
Katrina K Hoyer et al.
Immunological reviews, 226, 19-28 (2009-01-24)
Interleukin-2 (IL-2) has multiple, sometimes opposing, functions during an inflammatory response. It is a potent inducer of T-cell proliferation and T-helper 1 (Th1) and Th2 effector T-cell differentiation and provides T cells with a long-lasting competitive advantage resulting in the
Arian Laurence et al.
Immunity, 26(3), 371-381 (2007-03-17)
Recent work has identified a new subset of effector T cells that produces interleukin (IL)-17 known as T helper 17 (Th17) cells, which is involved in the pathophysiology of inflammatory diseases and is thought to be developmentally related to regulatory
Hans Dooms et al.
Journal of immunology (Baltimore, Md. : 1950), 172(10), 5973-5979 (2004-05-07)
The acquisition of long-term survival potential by activated T lymphocytes is essential to ensure the successful development of a memory population in the competitive environment of the lymphoid system. The factors that grant competitiveness for survival to primed T cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service