Skip to Content
Merck
All Photos(1)

Key Documents

I5027

Sigma-Aldrich

Anti-Interferon-γ antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IFN-γ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

immunohistochemistry: 5-15 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ifng(15978)
rat ... Ifng(25712)

Looking for similar products? Visit Product Comparison Guide

Immunogen

recombinant rat interferon-γ expressed in E. coli.

Biochem/physiol Actions

Interferons (IFNs) are cytokines that are secreted in response to viral or bacterial infections and tumorigenesis. IFN-γ binds to specific receptor complex consisting of IFNγR1 and IFNγR2. There are many pathways that are mediated by IFN-γ binding such as JAK/STAT1, AP-1, NF-κB, STAT3 and STAT5. IFN-γ is pleotropic and performs various functions related to immune response, inflammation, differentiation and activation of T cells, cell cycle and apoptosis.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huixia Chen et al.
Journal of biochemical and molecular toxicology, 38(7), e23761-e23761 (2024-07-02)
Non-small cell cancer (NSCLC) is the most common cancer in the world, but its effective therapeutic methods are limited. Tilianin and sufentanil alleviate various human tumors. This research aimed to clarify the functions and mechanisms of Tilianin and sufentanil in
Jingjie Wei et al.
BMC endocrine disorders, 22(1), 317-317 (2022-12-14)
IFN-γ is critical for both thyroid and ovarian function, while thyroxine, secreted from the thyroid gland, regulates the ovarian function via the hypothalamus-pituitary -ovary axis. However, the effect of thyroxine on INF-γ involved in the regulation of hypothalamic pituitary ovarian axis
Kate Schroder et al.
Journal of leukocyte biology, 75(2), 163-189 (2003-10-04)
Interferon-gamma (IFN-gamma) coordinates a diverse array of cellular programs through transcriptional regulation of immunologically relevant genes. This article reviews the current understanding of IFN-gamma ligand, receptor, signal transduction, and cellular effects with a focus on macrophage responses and to a
Leonidas C Platanias
Nature reviews. Immunology, 5(5), 375-386 (2005-05-03)
Interferons are cytokines that have antiviral, antiproliferative and immunomodulatory effects. Because of these important properties, in the past two decades, major research efforts have been undertaken to understand the signalling mechanisms through which these cytokines induce their effects. Since the
S E Ealick et al.
Science (New York, N.Y.), 252(5006), 698-702 (1991-05-03)
The x-ray crystal structure of recombinant human interferon-gamma has been determined with the use of multiple-isomorphous-replacement techniques. Interferon-gamma, which is dimeric in solution, crystallizes with two dimers related by a noncrystallographic twofold axis in the asymmetric unit. The protein is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service