Skip to Content
Merck

Skip To

F4883

Fibrinogen from human plasma

35-65% protein (≥90% of protein is clottable).

Synonym(s):

Factor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-598-6
NACRES:
NA.32
MDL number:
Form:
powder
Biological source:
human plasma
Mol wt:
α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

description

essentially plasmin(ogen) free

form

powder

quality

35-65% protein (≥90% of protein is clottable).

mol wt

α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa

concentration

35-65% protein (biuret)

technique(s)

cell culture | mammalian: suitable

solubility

0.9% NaCl: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Fibrinogen was the first blood coagulation factor to be identified, and it is a precursor of fibrin protein. This protein is composed of two chains, each of which is composed of three chains- α, β and γ. These three subunits are coded by three genes located in the human chromosomal locus 4q23-q32, and spans 51kb.

Application

Fibrinogen from human plasma has been used for-
  • fibrinogen adsorption assay performed on swCNTs (single-walled carbon nanotubes)
  • the characterization of fibrinogen structure in the presence of CNT using CD (Circular dichroism) spectroscopy and temperature scan
  • the preparation of RBC suspension obtained from venous blood of antecubital vein, to assess RBC aggregability
  • fibrinolysis assay performed on mouse embryonic fibroblasts (MEFs)
  • the preparation of fibrinogen-labeled gold (Fgn/Au)
  • the quantification of in vitro angiogenesis by human microvascular endothelial cells and
  • the assessment of fibrin-induced apoptosis and differentiation-regulation in trophoblast obtained from term human placentas.

Biochem/physiol Actions

Fibrinogen is an acute-phase inflammatory protein, which plays an essential role in blood clotting. Coronary heart disease is linked to the plasma levels of fibrinogen in Chinese population, and might have potential as a marker and risk factor for the same. A -455 allele of fibrinogen β (FGB) chain is linked with poor survival in Caucasian women with Finnish stroke. Ischemic stroke preceded by hyperfibrinogenemia has poor outcome, which is independent of the baseline fibrinogen concentration.
Acute phase protein that is part of the coagulation cascade of proteins.
Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Thrombin rapidly proteolyses fibrinogen, releasing fibrinopeptide A. The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules. Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel. The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII. The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding. The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

Features and Benefits

Source material has tested negative for HIV and HBsAg.

Physical form

Lyophilized powder containing sodium citrate and sodium chloride.

Analysis Note

Protein determined by biuret

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3879F6755341573
description

35-65% protein (≥90% of protein is clottable).

description

50-70% protein (≥80% of protein is clottable)

description

60-80% protein (≥60% of protein is clottable)

description

Fibrinogen, CAS 9001-32-5, is a soluble plasma glycoprotein that is synthesized in the hepatic parenchymal cells. Contains mpre than 95% clottable protein.

technique(s)

cell culture | mammalian: suitable

technique(s)

ELISA: suitable

technique(s)

cell culture | mammalian: suitable, immunoblotting: suitable

technique(s)

-

biological source

human plasma

biological source

human plasma

biological source

rat plasma

biological source

-

concentration

35-65% protein (biuret)

concentration

50-70% protein (biuret)

concentration

60-80% (biuret)

concentration

-

mol wt

α-chain 63.5 kDa, γ chain 47 kDa (about 4% carbohydrate content), β-chain 56 kDa, soluble dimer 340 kDa

mol wt

β-chain 56 kDa, α-chain 63.5 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa

mol wt

α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa

mol wt

-

form

powder

form

powder

form

powder

form

lyophilized

solubility

0.9% NaCl: soluble

solubility

0.9% NaCl: soluble 10 mg/mL

solubility

-

solubility

sterile distilled water: soluble


Still not finding the right product?

Explore all of our products under Fibrinogen from human plasma


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. storage and thawing Is it possible to store at -20 freezer and repeated freezing and thawing

    1 answer
    1. This product has not been tested for freeze-thaw stability. However, in powder form, repeatedly removing this product from a freezer for the purpose of preparing fresh solutions, is not expected to be detrimental to the protein. Prepared solutions may be stored up to 1 week at 2 - 8 °C. It is not recommended to freeze stock solutions, as the freeze-thaw process may lead to protein degradation.
      Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/299/301/f4883pis.pdf

      Helpful?

  2. F4883-500MG contains Sodium Citrate and Sodium Chlorure. Is it possible to have their concentration ? What is the volume of sodium chloride we have to put in the bottle please?

    1 answer
    1. This material is lyophilized from a solution of 75 mM NaCl (4.39 mg/mL), 10 mM Na Citrate (2.94 mg/mL). It can be solubilized in 0.85% (w/v) sodium chloride to 2 mg/ml, by layering the protein on top of the saline solution and placing it in a 37°C water bath for 4-6 hours, with gentle agitation.

      Helpful?

  3. Hello. For your F4883-500MG Fibrinogen product, is there 500mg of pure fibrinogen in the vial? Or is there 500 mg of reagent - of which a fraction is clottable Fibrinogen? Do you have preparation instructions for this reagent?

    1 answer
    1. F4883-500MG contains 500 mg of solid. Fibrinogen can be solubilized in 0.85% (w/v) sodium chloride (2 mg/ml) by layering the protein on top of the saline solution and placing it in a 37 °C water bath for 4-6 hours with gentle agitation.

      Helpful?

Reviews

No rating value

Active Filters