Skip to Content
Merck
All Photos(1)

Key Documents

I2406

Sigma-Aldrich

IL-11 human

>97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

IL-11

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Interleukin-11 human, >97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.06-0.24 ng/mL ED50

mol wt

23 kDa

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL11(3589)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-11 (IL-11) is synthesized by many mesenchymal origin cells like keratinocytes, synoviocytes, fibroblasts, bone marrow stromal cells, osteoblasts, and chondrocytes.
The gene IL-11 (interleukin 11) is mapped to human chromosome 19q13.4. It is a secreted protein.

Biochem/physiol Actions

Interleukin 11 (IL-11) is involved in promoting tissue fibrosis and regulates the function of chondrocytes, synoviocytes, and B cells. It is also involved in bone metabolism and tissue remodeling.
Interleukin-11, also called adipogenesis inhibitory factor (AGIF), is a pleotropic cytokine that acts on hematopoietic progenitor cells and stromal cells. Although recombinant human IL-11 has a predicted molecular mass of 19 kDa, it migrates as a 23 kDa band in SDS-PAGE. It is a functional homolog of IL-6, inducing the proliferation of certain plasmacytoma cell lines and acute phase protein secretion in the liver. IL-11 stimulates the production of erythrocytes, megakaryocytes, and T cell development of antibody producing B cells.

Physical form

Lyophilized from a 0.2 μm filtered solution of PBS and EDTA with BSA as a carrier protein.

Analysis Note

The biological activity is measured in a cell proliferation assay using T11, a subline of the IL-6-dependent murine plasmacytoma cell line T1165.85.2.1 that has been adapted to grow in IL-11.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Lebeau et al.
FEBS letters, 407(2), 141-147 (1997-04-28)
Long-term stable Ba/F3 transfectants (B13R alpha1 and B13R alpha2) expressing two isoforms of the human IL-IIR alpha receptor (alpha1 full length or alpha2 lacking the cytoplasmic domain) in combination with human gp130 were established. IL-11R alpha1 and IL-11R alpha2 were
I Kawashima et al.
FEBS letters, 283(2), 199-202 (1991-06-03)
A cDNA encoding a novel adipogenesis inhibitory factor (AGIF) that inhibits the process of adipogenesis in mouse 3T3-L1 preadipocytes was cloned from a cDNA library of the human bone marrow-derived stromal cell line KM-102. The cloned cDNA contains an open
V F Quesniaux et al.
Blood, 80(5), 1218-1223 (1992-09-01)
Interleukin-11 (IL-11), a pleiotropic cytokine originally isolated from a primate bone marrow stromal cell line, has been shown to stimulate T-cell-dependent B-cell maturation, megakaryopoiesis, and various stages of myeloid differentiation, but to inhibit adipogenesis. Because stromal cells are essential for
W Klein et al.
Genes and immunity, 3(8), 494-496 (2002-12-18)
Inflammatory bowel diseases (IBD) are multifactorial disorders characterised by the host's inability to limit the inflammatory response to luminal antigens. The association of polymorphisms in the CARD15 gene with Crohn's disease (CD) demonstrates the relevance of activated transcription factor NF(kappa)B
S X Leng et al.
The international journal of biochemistry & cell biology, 29(8-9), 1059-1062 (1997-08-01)
Interleukin-11 (IL-11) is an IL-6-type cytokine that is produced by a variety of stromal cells including fibroblasts, epithelial cells and osteoblasts. It binds to a multimeric receptor complex which contains an IL-11-specific alpha subunit and a promiscuous 130 kDa beta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service