Skip to Content
Merck
All Photos(2)

Key Documents

I1771

Sigma-Aldrich

Interleukin-13 human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-13

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

<10.000 ng/mL ED50

mol wt

12 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL13(3596)

Biochem/physiol Actions

IL-13 is known to be overexpressed in glioblastoma tumors and cell lines. Elevated IL-13 expression is also observed in parasitic infections. It stimulates fibrosis in many infectious diseases, and acts as a profibrogenic factor.
Interleukin-13 is a pleiotropic cytokine produced by activated Th2 cells in the mouse and human. IL-13 is secreted mainly as an unglycosylated protein of 132 amino acids with a molecular weight of 10,000 daltons. A 4.3kb DNA fragment of the mouse IL-13 gene was sequenced and occurs as a single copy mapping a chromosome 11. In the human, a 4.6 kb DNA segment of the IL-13 gene occurs as a single copy and maps to chromosome 5. IL-13 induces B cell proliferation and also induces IgE switching. The proliferative activity of human and mouse IL-13 products are tested in culture using the human erythroleukemic cell line TF-1.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 250 μg BSA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Yun Lee et al.
Journal of immunology (Baltimore, Md. : 1950), 194(12), 5990-5998 (2015-05-15)
Mucin overproduction is a hallmark of otitis media (OM). Streptococcus pneumoniae is one of the most common bacterial pathogens causing OM. Mucin MUC5AC plays an important role in mucociliary clearance of bacterial pathogens. However, if uncontrolled, excessive mucus contributes significantly
Gui Yang et al.
The Journal of biological chemistry, 290(20), 12858-12867 (2015-04-04)
Restoration of the antigen (Ag)-specific immune tolerance in an allergic environment is refractory. B cells are involved in immune regulation. Whether B cells facilitate the generation of Ag-specific immune tolerance in an allergic environment requires further investigation. This paper aims
An IL-13 promoter polymorphism associated with liver fibrosis in patients with Schistosoma japonicum.
Long X et al
PLoS ONE, 10(8), e0135360-e0135360 (2015)
Qi Liu et al.
The Journal of heart valve disease, 24(4), 496-501 (2016-02-24)
Interleukin (IL)-13 is a major inducer of fibrosis in many chronic infectious diseases, yet few studies have reported its role in valvular fibrosis in patients with rheumatic heart disease (RHD). The study aim was to investigate the role of IL-13
Kihyuk Shin et al.
Clinical and experimental medicine, 15(3), 233-244 (2014-06-14)
The leukotrienes (LTs) enhance allergen- and interleukin (IL)-13-dependent allergic lung inflammatory disease. However, the precise requirement of LTs and the mechanism by which they elicit allergic lung responses remain uncertain. To clarify the involvement of LTs in respiratory allergen- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service