Skip to Content
Merck
All Photos(1)

Key Documents

C2020

Sigma-Aldrich

α-Cyano-4-hydroxycinnamic acid

≥98% (TLC), powder, monocarboxylic acid transport inhibitor

Synonym(s):

α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=C(CN)CO2H
CAS Number:
Molecular Weight:
189.17
Beilstein:
3271427
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder

Assay

≥98% (TLC)

form

powder

color

yellow

mp

245-250 °C (lit.)

solubility

H2O: slightly soluble
methanol: water: soluble
polar organic solvents: soluble

storage temp.

2-8°C

SMILES string

OC(=O)\C(=C\c1ccc(O)cc1)C#N

InChI

1S/C10H7NO3/c11-6-8(10(13)14)5-7-1-3-9(12)4-2-7/h1-5,12H,(H,13,14)/b8-5+

InChI key

AFVLVVWMAFSXCK-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Cyano-4-hydroxycinnamic acid has been used to block monocarboxylate transporters.
α-Cyano-4-hydroxycinnamic acid is a useful hydrophobic matrix solution for matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry. Antibiotics, peptide nucleic acids (a new class of DNA mimics), and proteins with masses as high as 66,000 Da have been successfully analyzed by using this as a matrix solution.

Biochem/physiol Actions

α-Cyano-4-hydroxycinnamic acid acts as a specific inhibitor of monocarboxylic acid transport, including lactate and pyruvate transport. It is also reported to block β-cell apical anion exchange (IC50 of 2.4 mM).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Emmons
The American journal of physiology, 276(4 Pt 2), F635-F643 (1999-04-13)
To functionally characterize transport properties of the apical anion exchanger of rabbit beta-intercalated cells, the mean change in anion exchange activity, dpHi/dt (where pHi is intracellular pH), was measured in response to lumen Cl- replacement with gluconate in perfused cortical
Elizabeth Csaszar et al.
Biotechnology and bioengineering, 103(2), 402-412 (2009-03-07)
An automated delivery system for cell culture applications would permit studying more complex culture strategies and simplify measures taken to expose cells to unstable molecules. We are interested in understanding how intracellular TAT-HOXB4 protein concentration affects hematopoietic stem cell (HSC)
Adaptive cellular mechanisms in response to Glutamine-starvation
Eliasen MM, et al.
Frontiers in Bioscience, 11, 3199-3211 (2006)
U Schneider et al.
Neuroscience, 53(4), 1153-1162 (1993-04-01)
Exposure of nervous tissue to hypoxia results in interstitial acidification. There is evidence for concomitant decrease in extracellular pH to the increase in tissue lactate. In the present study, we used double-barrelled pH-sensitive microelectrodes to investigate the link between lactate
E T Sze et al.
Journal of the American Society for Mass Spectrometry, 9(2), 166-174 (1998-07-29)
We report a simple method for converting solid matrices into useful matrix solutions for matrix-assisted laser desorption/ionization (MALDI). This method is based on the dissolution of the solid matrix in a liquid support of low volatility such as glycerol. An

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service