Recommended Products
form
solution
Quality Level
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
concentration
0.6%
pH
8.5
application(s)
hematology
histology
storage temp.
room temp
Application
Intended for use in Sigma Gram Stain Procedure, HT90.
Components
Certified safranin 0.6% in 20% ethyl alcohol, SD3A.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
100.0 °F - closed cup
Flash Point(C)
37.8 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biomaterials, 177, 1-13 (2018-06-09)
Overexpression and knockdown of specific proteins can control stem cell differentiation for therapeutic purposes. In this study, we fabricated RUNX2, SOX9, and C/EBPα plasmid DNAs (pDNAs) and ATF4-targeting shRNA (shATF4) to induce osteogenesis, chondrogenesis, and adipogenesis of human mesenchymal stem
Inhibition of integrative repair of the meniscus following acute exposure to interleukin-1 in vitro.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 26(4), 504-512 (2007-12-01)
Damage or loss of the meniscus is associated with progressive osteoarthritic degeneration of the knee joint. Injured and degenerative joints are characterized by elevated levels of the pro-inflammatory cytokine interleukin-1 (IL-1), which with prolonged exposure can induce catabolic and anti-anabolic
Arthritis & rheumatology (Hoboken, N.J.), 71(4), 612-625 (2018-11-06)
It remains unclear if and how inflammation and new bone formation in spondyloarthritis (SpA) are coupled. We undertook this study to assess the hypothesis that interleukin-17A (IL-17A) is a pivotal driver of both processes. The effect of tumor necrosis factor
International journal of molecular sciences, 21(19) (2020-10-03)
Discoidin domain receptor 1 (Drd1) is a collagen-binding membrane protein, but its role in osteoblasts during osteogenesis remains undefined. We generated inducible osteoblast-specific Ddr1 knockout (OKOΔDdr1) mice; their stature at birth, body weight and body length were significantly decreased compared
Materials science & engineering. C, Materials for biological applications, 109, 110578-110578 (2020-04-02)
The current gold standard for nasal reconstruction after rhinectomy or severe trauma includes transposition of autologous cartilage grafts in conjunction with coverage using an autologous skin flap. Harvesting autologous cartilage requires a major additional procedure that may create donor site
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service