Skip to Content
Merck
All Photos(6)

Key Documents

HPA037609

Sigma-Aldrich

Anti-LOXL4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ21889, Anti-LOXC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RRHRGYLSETVSNALGPQGRRLEEVRLKPILASAKQHSPVTEGAVEVKYEGHWRQVCDQGWTMN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LOXL4(84171)

General description

Lysyl oxidase like 4 (LOXL4) is a copper-dependent amine oxidase and the LOXL4 gene is mapped to human chromosome 10q24.2. It is expressed in the pancreas and testes. LOXL4 comprises an N-terminal signal peptide and four scavenger receptor domains.

Immunogen

lysyl oxidase-like 4

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Lysyl oxidase like 4 (LOXL4) scavenger receptor cysteine-rich (SRCR) regions are critical for interaction with ligands. LOXL4 is implicated in particular with the head and neck squamous cell carcinomas, esophageal squamous cell carcinoma (ESCC) as well as with diverse cancer types. However, LOXL4 gene displays low expression in liver and bladder cancer. LOXL4 protein is crucial for extracellular matrix (ECM) homeostasis and mediates the oxidative deamination in proteins like collagen and elastin at their lysine residues. Its expression levels also modulate the ECM remodeling. Studies have highlighted elevated expression of LOXL4 in liver cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80265

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijie Xie et al.
Amino acids, 51(5), 813-828 (2019-03-23)
Lysyl oxidase-like 4 (LOXL4), a member of the LOX family proteins, catalyzes oxidative deamination of lysine residues in collagen and elastin, which are responsible for maintaining extracellular matrix homeostasis. In this study, the mRNA expression of LOXL4 in seven esophageal
Oscar Busnadiego et al.
Molecular and cellular biology, 33(12), 2388-2401 (2013-04-11)
Transforming growth factor β1 (TGF-β1) is a pleiotropic factor involved in the regulation of extracellular matrix (ECM) synthesis and remodeling. In search for novel genes mediating the action of TGF-β1 on vascular ECM, we identified the member of the lysyl
Tibor Görögh et al.
International journal of oncology, 33(5), 1091-1098 (2008-10-25)
Lysyl oxidases are a family of five copper-dependent amine oxidases including LOX, LOXL, LOXL2, LOXL3 and LOXL4. LOX and LOXL are essential for the assembly and maintenance of extracellular matrixes. LOXL2, LOXL3 and LOXL4, secreted and active enzymes, were also
Vincent Palmieri et al.
The Journal of pathology, 251(2), 213-223 (2020-04-17)
Colorectal cancer liver metastases (CRCLM) that present with a replacement histopathological growth pattern (HGP) are resistant to neoadjuvant anti-angiogenic therapy. Surrogate biomarkers are not available to preoperatively identify patients with these tumors. Here we identify differentially expressed genes between CRCLM
Rongkun Li et al.
Molecular cancer, 18(1), 18-18 (2019-02-02)
Lysyl oxidase-like 4 (LOXL4) has been found to be dysregulated in several human malignancies, including hepatocellular carcinoma (HCC). However, the role of LOXL4 in HCC progression remains largely unclear. In this study, we investigated the clinical significance and biological involvement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service