Skip to Content
Merck
All Photos(2)

Key Documents

HPA038161

Sigma-Aldrich

Anti-CEP83 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CCDC41, Anti-NY-REN-58

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LKRLQEKVEVLEAKKEELETENQVLNRQNVPFEDYTRLQKRLKDIQRRHNEFRSLILVPNMPPTASINPVSFQSSAMVPSMELPFPPHMQEEQHQRELSL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCDC41(51134)

General description

The gene CEP83 (centrosomal protein 83) encodes a protein with coiled-coil domains.

Immunogen

centrosomal protein 83kDa

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CEP83 antibody produced in rabbit has been used in western blotting and immunofluorescence.

Biochem/physiol Actions

CEP83 (centrosomal protein 83) plays an important role in ciliogenesis. It is part of DAPs (distal appendages) in centrioles. In ciliogenesis, DAPs are needed for docking and attachment of the mother centriole to the cellular surface. CEP83 is also required for bringing other components (for instance CEP164) to the mother centriole. Mutation in this gene is associated with early-onset infantile nephronophthisis and intellectual disability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81162

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren T Evans et al.
The EMBO journal, 40(4), e105106-e105106 (2020-12-23)
Centriole copy number is tightly maintained by the once-per-cycle duplication of these organelles. Centrioles constitute the core of centrosomes, which organize the microtubule cytoskeleton and form the poles of the mitotic spindle. Centrosome amplification is frequently observed in tumors, where
Rannar Airik et al.
PloS one, 11(5), e0156081-e0156081 (2016-05-26)
Recessive mutations in the SDCCAG8 gene cause a nephronophthisis-related ciliopathy with Bardet-Biedl syndrome-like features in humans. Our previous characterization of the orthologous Sdccag8gt/gt mouse model recapitulated the retinal-renal disease phenotypes and identified impaired DNA damage response signaling as an underlying
Bahtiyar Kurtulmus et al.
Journal of cell science, 131(18) (2018-08-23)
Cilia perform essential signalling functions during development and tissue homeostasis. A key event in ciliogenesis occurs when the distal appendages of the mother centriole form a platform that docks ciliary vesicles and removes CP110-Cep97 inhibitory complexes. Here, we analysed the
Dong Kong et al.
The Journal of cell biology, 206(7), 855-865 (2014-09-24)
Newly formed centrioles in cycling cells undergo a maturation process that is almost two cell cycles long before they become competent to function as microtubule-organizing centers and basal bodies. As a result, each cell contains three generations of centrioles, only
Chuan Chen et al.
Science advances, 7(26) (2021-06-25)
Mutation of ciliopathy protein HYLS1 causes the perinatal lethal hydrolethalus syndrome (HLS), yet the underlying molecular etiology and pathogenesis remain elusive. Here, we reveal unexpected mechanistic insights into the role of mammalian HYLS1 in regulating primary cilia. HYLS1 is recruited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service