Skip to Content
Merck
All Photos(1)

Key Documents

HPA019547

Sigma-Aldrich

Anti-LMBRD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HDAg-L-interacting protein NESI, Anti-LMBR1 domain-containing protein 1, Anti-Nuclear export signal-interacting protein, Anti-Probable lysosomal cobalamin transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KGTRSAAYERLENTEDIEEVEQHIQTIKSKSKDGRPLPARDKRALKQFEERLRTLKKRERHLEFIENSWWTKFCGALRP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMBRD1(55788)

General description

LMBRD1 (LMBR1 domain containing 1) is an integral lysosomal membrane protein located on chromosome 6q13. It is expressed in human liver tissue, and it comprises a putative actin-binding site and a bipartite nuclear localization signal.

Immunogen

Probable lysosomal cobalamin transporter recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

LMBRD1 (LMBR1 domain containing 1) may be involved in the lysosomal cobalamin transport. It was found to mediate transport of cobalamin through the lysosomal membranes. Mutation in LMBRD1 gene causes the cblF (Cobalamin F defect) defect of vitamin B12 (cobalamin) metabolism, characterized with accumulation of internalized free cobalamin in the lysosome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73290

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insa Buers et al.
Journal of cellular and molecular medicine, 20(8), 1523-1533 (2016-04-12)
The rare inborn cblF defect of cobalamin metabolism is caused by mutations in the limb region 1 (LMBR1) domain containing 1 gene (LMBRD1). This defect is characterized by massive accumulation of free cobalamin in lysosomes and loss of mitochondrial succinyl-CoA
Yun-Hsin Wang et al.
Journal of virology, 79(13), 8113-8120 (2005-06-16)
The process of host factor-mediated nucleocytoplasmic transport is critical for diverse cellular events in eukaryotes and the life cycle of viruses. We have previously identified a chromosome region maintenance 1-independent nuclear export signal (NES) at the C terminus of the
Frank Rutsch et al.
Nature genetics, 41(2), 234-239 (2009-01-13)
Vitamin B(12) (cobalamin) is essential in animals for metabolism of branched chain amino acids and odd chain fatty acids, and for remethylation of homocysteine to methionine. In the cblF inborn error of vitamin B(12) metabolism, free vitamin accumulates in lysosomes
Susann Gailus et al.
Journal of inherited metabolic disease, 33(1), 17-24 (2010-02-04)
In the cblF defect of vitamin B(12) (cobalamin) metabolism, cobalamin is trapped in lysosomes. Consequently, cobalamin coenzyme synthesis is blocked, and cofactors for methionine synthase and methylmalonyl-coenzyme A (CoA) mutase are deficient. We recently identified LMBRD1 as the causative gene
Isabelle R Miousse et al.
Molecular genetics and metabolism, 102(4), 505-507 (2011-02-10)
The cblF disorder, characterized by accumulation of internalized cobalamin in the lysosome, is caused by mutations in the LMBRD1 gene which encodes an integral lysosomal membrane protein. We describe novel mutations in LMBRD1 in three patients: two splice site mutations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service