Skip to Content
Merck
All Photos(10)

Key Documents

ZRB06711

Sigma-Aldrich

Anti-FADD Antibody, clone 11N7, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

FAS-associated death domain protein, FAS-associating death domain-containing protein, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, Protein FADD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11N7, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 23.28 kDa
observed mol wt ~26 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

gene knockdown: suitable using 1:250
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 11N7 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human FAS-associated death domain protein (FADD).

Immunogen

His-tagged full-length recombinant human FAS-associated death domain protein (FADD).

Application

Anti-FADD, clone 11N7, ZooMAb, Cat. No. Cat. No. ZRB06711, is a recombinant Rabbit Monoclonal antibody that detects human FAS-associated death domain and has been tested for use in Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected FADD in MCF7 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FADD in human kidney tissue sections.

RNAi/siRNA/Gene Knockdown Analysis: A 1:250 dilution from a representative lot detected FADD lysates from Wild type HeLa cells, but not in FADD knockout HeLa cells.

Target description

FAS-associated death domain protein (UniProt: Q13158; also known as FAS-associating death domain-containing protein, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, Protein FADD) is encoded by the FADD (also known as MORT1, GIG3) gene (Gene ID: 8772) in human. FADD is an apoptotic adaptor molecule that bridges death receptor signaling to the caspase cascade. It is expressed in a wide variety of tissues, except for peripheral blood mononuclear leukocytes. FADD contains a death domain at its C-terminus and a death effector domain (DED) at its N-terminal. The death domain is responsible for FADD binding to the activated trimerized Fas receptor. FADD recruits procaspase-8 through its N-terminal DED, which is then activated following the cleavage of pro domain. The death inducing signaling complex (DISC) formed by Fas receptor, FADD, and caspase-8 is essential in triggering apoptotic events in cells. Human FADD is phosphorylated mainly at Ser194. Defects in FADD gene have been linked to recurrent infections, hepatic dysfunction, and cardiovascular malformations. These subjects display high circulating levels of CD4(-) and CD8(-) cells and elevated levels of IL-10. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service