Skip to Content
Merck
All Photos(9)

Key Documents

HPA018425

Sigma-Aldrich

Anti-G3BP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-G3BP-2, Anti-GAP SH3 domain-binding protein 2, Anti-Ras GTPase-activating protein-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

KNLEELEEKSTTPPPAEPVSLPQEPPKPRVEAKPEVQSQPPRVREQRPRERPGFPPRGPRPGRGDMEQNDS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G3BP2(9908)

General description

The gene G3BP2 (GAP SH3 domain-binding protein-2) has been mapped to human chromosome 4q21.1. G3BP2 is ubiquitously expressed and is mainly present in the cytoplasm. However, it is capable of shuttling into the nucleus in cell-cycle dependent manner. G3BP2 include an NTF2 (nuclear transport factor)-like domain and two RNA-binding motifs.

Immunogen

Ras GTPase-activating protein-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-G3BP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GAP SH3 domain-binding protein-2 (G3BP2) is identified as a gene for predicting the presence of lymph node metastasis in primary oral squamous cell carcinoma. G3BP2 is a negative modulator of p53 function. Depletion of G3BP2 leads to up-regulation of p53 and its overexpression leads to the cytoplasmic redistribution of p53. G3BP2 binds mechanomediator TWIST1 in the cytoplasm. Loss of G3BP2 leads to nuclear localization of TWIST1 which in response to biomechanical signals induce epithelial-mesenchymal transition, promoting tumor invasion and metastasis. Overexpression of G3BP2 induces formation of cytoplasmic RNA granules called stress granules (SGs). Knockdown of G3BP2 reduces the number of SG-positive cells. During Semliki Forest virus infection, the C-terminal domain of the viral nonstructural protein-3 forms a complex with G3BP2 and inhibits the formation of SGs on viral mRNAs, thus impairing antiviral defense.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73970

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weina Wang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 11(6), e2305068-e2305068 (2023-12-13)
Primary cilia are conserved organelles in most mammalian cells, acting as "antennae" to sense external signals. Maintaining a physiological cilium length is required for cilium function. MicroRNAs (miRNAs) are potent gene expression regulators, and aberrant miRNA expression is closely associated
M M Kim et al.
Oncogene, 26(29), 4209-4215 (2007-02-14)
Inactivation of the p53 tumor suppressor pathway is a critical step in human tumorigenesis. In addition to mutations, p53 can be functionally silenced through its increased degradation, inhibition of its transcriptional activity and/or its inappropriate subcellular localization. Using a proteomic
Ying Sun et al.
Oncogene, 38(24), 4856-4874 (2019-02-26)
Identification of molecular alterations driving breast cancer progression is critical for the development of effective therapy. In this study, we show that the level of α-parvin is elevated in triple-negative breast cancer cells. The depletion of α-parvin from triple-negative breast
Marc D Panas et al.
Molecular biology of the cell, 23(24), 4701-4712 (2012-10-23)
Dynamic, mRNA-containing stress granules (SGs) form in the cytoplasm of cells under environmental stresses, including viral infection. Many viruses appear to employ mechanisms to disrupt the formation of SGs on their mRNAs, suggesting that they represent a cellular defense against
Amy E Rose et al.
Cancer research, 71(7), 2561-2571 (2011-02-24)
Superficial spreading melanoma (SSM) and nodular melanoma (NM) are believed to represent sequential phases of linear progression from radial to vertical growth. Several lines of clinical, pathologic, and epidemiologic evidence suggest, however, that SSM and NM might be the result

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service