Skip to Content
Merck
All Photos(3)

Documents

761605

Sigma-Aldrich

BrettPhos Pd G3

98%

Synonym(s):

[(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulfonate methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H66NO5PPdS
CAS Number:
Molecular Weight:
906.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reagent type: catalyst
core: palladium
reaction type: Cross Couplings

mp

150-193 °C (decomposition)

functional group

amine

SMILES string

CS(=O)(O[Pd]c1c(c2c(N)cccc2)cccc1)=O.COc3c(P(C4CCCCC4)C5CCCCC5)c(c6c(C(C)C)cc(C(C)C)cc6C(C)C)c(OC)cc3

InChI

1S/C35H53O2P.C12H10N.CH4O3S.Pd/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h19-25,27-28H,9-18H2,1-8H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

PQYBJDCHLVJYSD-UHFFFAOYSA-M

General description

BrettPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. BrettPhos Pd G3 is an excellent reagent for Buchwald-Hartwig cross-coupling reaction. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

BrettPhos Pd G3 may be used for the C-N coupling reaction between monosubstituted ureas and 1,2 dihaloaromatic system to form benzimidazolones.
It may be used in the synthesis of following compounds:
  • [Pd(cinnamyl)(BrettPhos)]OTf by reacting with [(cinnamyl)PdCl]2 and AgOTf.
  • [Pd(crotyl)(BrettPhos)]OTf by reacting with [(crotyl)PdCl]2 and AgOTf.
  • [Pd(allyl)(BrettPhos)]OTf by reacting with [(allyl)PdCl]2 and AgOTf.
  • Pd(allyl)(BrettPhos)Clby reacting with [(allyl)PdCl]2.
For small scale and high throughput uses, product is also available as ChemBeads (928321)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective Synthesis of Benzimidazolones via Cascade C-N Coupling of Monosubstituted Ureas.
Ernst JB, et al.
Organic Letters, 16(14), 3844-3846 (2014)
Generating Active ?L-Pd (0)? via Neutral or Cationic Π-Allylpalladium Complexes Featuring Biaryl/Bipyrazolylphosphines: Synthetic, Mechanistic, and Structure-Activity Studies in Challenging Cross-Coupling Reactions.
DeAngelis AJ, et al.
The Journal of Organic Chemistry, 80(13), 6794-6813 (2015)
2-Aminobiphenyl Palladacycles: The ?Most Powerful? Precatalysts in C-C and C-Heteroatom Cross-Couplings.
Bruneau A, et al.
ACS Catalysis, 5(2), 1386-1396 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service