Skip to Content
Merck
All Photos(6)

Key Documents

HPA017880

Sigma-Aldrich

Anti-TOX4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Epidermal Langerhans cell protein LCP1, Anti-TOX high mobility group box family member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MQQPPPQKVRINLQQQPPPLQIKSVPLPTLKMQTTLVPPTVESSPERPMNNSPEAHTVEAPSPETICEMITDVVPEVESPSQMDVELVSGSPVALSPQPRCVRSGCENPPIVSKDWDNEYCSNECVVKHCRDVFLAWVAS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOX4(9878)

General description

TOX high mobility group box family member 4 (TOX4) belongs to the HMG-box protein family and is expressed in the nucleus. It possesses lysine-based NLS (nuclear localization signal) and DNA-binding HMG box motif.

Immunogen

TOX high mobility group box family member 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-TOX4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-TOX4 antibody produced in rabbit has been used for epifluorescence microscopy.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

TOX high mobility group box family member 4 (TOX4) recognizes DNA adducts which are produced by platinum anticancer drugs and is suggested to be involved in cellular mechanisms against DNA damage. The carboxyl-terminal of TOX4 interacts with the amino-terminal region of protein phosphatase-1 nuclear targeting subunit (PNUTS) and this complex may play a role in the transcriptional regulation of genes. TOX4 also interacts with lens epithelium derived growth factor (LEDGF).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70391

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Langerhans cell protein 1 (LCP1) binds to PNUTS in the nucleus: implications for this complex in transcriptional regulation.
SJ Lee
Experimental & Molecular Medicine, 41(3), 189-200 (2009)
Mehdi Morchikh et al.
PloS one, 8(11), e81217-e81217 (2013-12-07)
PWWP domains are involved in the chromatin attachment of several proteins. They bind to both DNA and proteins and their interaction with specific histone methylation marks define them as a new class of histone code readers. The lens epithelium derived
Christophe Bounaix Morand du Puch et al.
Archives of biochemistry and biophysics, 507(2), 296-303 (2010-12-28)
Platinating agents are commonly prescribed anticancer drugs damaging DNA. Induced lesions are recognized by a wide range of proteins. These are involved in cellular mechanisms such as DNA repair, mediation of cytotoxicity or chromatin remodeling. They therefore constitute crucial actors
Li Ding et al.
Cell systems, 1(2), 141-151 (2016-05-03)
We combine a genome-scale RNAi screen in mouse epiblast stem cells (EpiSCs) with genetic interaction, protein localization, and "protein-level dependency" studies-a systematic technique that uncovers post-transcriptional regulation-to delineate the network of factors that control the expression of Oct4, a key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service