Skip to Content
Merck
All Photos(1)

Key Documents

HPA017324

Sigma-Aldrich

Anti-VIP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Peptide histidine methioninamide 27, Anti-VIP, Anti-VIP peptides, Anti-Vasoactive intestinal polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

EGANEPDQVSLKEDIDMLQNALAENDTPYYDVSRNARHADGVFTSDFSKLLGQLSAKKYLESLMGKRVSSNISEDPVPVKRHSDAVFTDNYTRLRKQMAVKKYLNSILNGKRSSEGESPDFPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIP(7432)

General description

VIP (vasoactive intestinal peptide) is a 28-amino acid peptide expressed in gastrointestinal tissues and neural tissues. It is originally isolated from porcine duodenum. VIP gene is mapped to human chromosome 6q25.2.

Immunogen

VIP peptides Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-VIP antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

VIP (vasoactive intestinal peptide) is involved in the VIP biosynthetic pathways. It also acts as a neurotransmitter in neural tissues and works in the relaxation of smooth muscle, stimulation of intestinal water and electrolyte secretion and release of insulin, glucagon and several anterior pituitary hormones. It participates in vasodilation. It acts as a potent agonist of the calcitonin receptor CALCR, with similar efficacy as calcitonin.  

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72428

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleni Angelopoulou et al.
Neuropeptides, 92, 102224-102224 (2022-01-09)
In female mammals, reproductive senescence is a complex process involving progressive ovarian dysfunction, associated with altered central control of the hypothalamic-pituitary-gonadal axis and desynchronization of the circadian system. The objective of this study was to investigate age-dependent changes in the
Camelia-Maria Monoranu et al.
Cancer genetics and cytogenetics, 182(1), 18-26 (2008-03-11)
Losses and rearrangements of genetic material on chromosome 6q are frequently found in several human malignancies, including primary central nervous system tumors. We previously used microsatellite analysis of ependymomas to identify frequent deletions in regions 6q15 approximately q16, 6q21 approximately
Jian-Nong Ma et al.
Biochemical pharmacology, 67(7), 1279-1284 (2004-03-12)
Many naturally occurring peptides exhibit a high degree of promiscuity across G-protein coupled receptor subtypes. The degree to which this phenomenon occurs, and its physiological significance is not well characterized. In addition, many 'orphan' peptides exist for which there are
Lieselot Dedeene et al.
Acta neuropathologica communications, 7(1), 189-189 (2019-12-04)
Motor-, behavior- and/or cognition-related symptoms are key hallmarks in patients with amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD) with TDP-43 pathology (FTLD-TDP), respectively. It has been reported that these patients also experience sleep disturbances, which might implicate a
Vania Carmona-Alcocer et al.
Frontiers in neuroscience, 17, 1177458-1177458 (2023-06-05)
Neuropeptide signaling modulates the function of central clock neurons in the suprachiasmatic nucleus (SCN) during development and adulthood. Arginine vasopressin (AVP) and vasoactive intestinal peptide (VIP) are expressed early in SCN development, but the precise timing of transcriptional onset has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service