Merck
  • Circadian sleep/wake-associated cells show dipeptide repeat protein aggregates in C9orf72-related ALS and FTLD cases.
Circadian sleep/wake-associated cells show dipeptide repeat protein aggregates in C9orf72-related ALS and FTLD cases.
Acta neuropathologica communications (2019-12-04)
Lieselot Dedeene, Evelien Van Schoor, Rik Vandenberghe, Philip Van Damme, Koen Poesen, Dietmar Rudolf Thal
ABSTRACT
Motor-, behavior- and/or cognition-related symptoms are key hallmarks in patients with amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD) with TDP-43 pathology (FTLD-TDP), respectively. It has been reported that these patients also experience sleep disturbances, which might implicate a disturbed circadian rhythm of the sleep/wake cycle. It remains unknown, however, whether cells involved in the circadian sleep/wake cycle are affected by ALS- and FTLD-related neuropathological changes including phosphorylated TDP-43 (pTDP-43) aggregates and dipeptide repeat protein (DPR) inclusions resulting from the C9orf72 hexanucleotide repeat expansion. Immunohistochemistry for DPR and pTDP-43 pathology was performed in post-mortem hypothalamus and pineal gland tissue of patients with ALS and/or FTLD-TDP with and without the C9orf72 repeat expansion and healthy controls. Circadian sleep/wake-associated cells, including pinealocytes and hypothalamic neurons related to the suprachiasmatic nucleus (SCN), were microscopically assessed. We observed numerous DPR inclusions (poly(GA), poly(GP), poly(GR) and poly(PR)) in the pinealocytes and few poly(GA) inclusions in the SCN-related neurons in C9orf72-related ALS and/or FTLD-TDP cases. These circadian sleep/wake-associated cells, however, were devoid of pTDP-43 pathology both in C9orf72- and nonC9orf72-related ALS and/or FTLD-TDP cases. Our neuropathological findings show that pinealocytes and, to a lesser extent, SCN-related neurons are affected by DPR pathology. This may reflect an involvement of these cells in sleep/wake disturbances observed in ALS and/or FTLD-TDP patients.
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GR) Antibody, clone 5A2, clone 5A2, 1 mg/mL, from rat
Sigma-Aldrich
Anti-VIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GA) Antibody, clone 5E9, clone 5E9, from mouse