Skip to Content
Merck
All Photos(4)

Key Documents

HPA010817

Sigma-Aldrich

Anti-FXYD5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dysadherin, Anti-FXYD domain-containing ion transport regulator 5 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

DTTSSSSADSTIMDIQVPTRAPDAVYTELQPTSPTPTWPADETPQPQTQTQQLEGTDGPLVTDPETHKSTKAAHPTDDTTTLSERPSPSTDVQTDPQTLKPSGFHEDDPFFYDEHTLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FXYD5(53827)

General description

FXYD5 (FXYD domain containing ion transport regulator 5) is a cancer-associated glycoprotein localized to the cell surface. It belongs to the family of FXYD proteins, which interacts with and modulate Na,K-ATPase. These proteins span the cell membrane once. The cDNA for this gene codes for a protein of 178 amino acids, containing a single transmembrane domain, a short cytoplasmic tail. It also has a putative O-glycosylated extracellular region as well as a signal peptide. This gene is present with the human chromosome locus 19q13.12. Under normal conditions, it is expressed in only specific tissues. These are generally epithelial tissues such as, lung, kidney and intestine. It is also expressed in normal lymphocytes, as well as in stratified squamous epithelia, in the basal and endothelial cells.

Immunogen

FXYD domain-containing ion transport regulator 5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FXYD5 (FXYD domain containing ion transport regulator 5) is responsible for transcriptionally down-regulating E-cadherin. Therefore, it facilitates tumor invasiveness and metastasis. It is suggested that FXYD5 interacts with actin, and has a role in the formation of microvilli. It is expressed in a wide range of cancer, such as breast, stomach, pancreatic and colon. It is also expressed in head and neck squamous cell carcinoma. It is also expressed in testis, epididymis and ejaculated sperms. In the ejaculated spermatozoa, it co-localizes with E-cadherin and Na,K-ATPase, and therefore, might be a regulator of sperm function. FXYD5 might have potential to determine the prognosis of disease-free survival in stage I NSCLC patients. Its expression level correlates to the risk level of gastrointestinal stromal tumors (GISTs), and thus, may be used for the prognosis of GISTs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71700

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia L Brazee et al.
Frontiers in immunology, 8, 623-623 (2017-06-18)
The alveolar epithelium secretes cytokines and chemokines that recruit immune cells to the lungs, which is essential for fighting infections but in excess can promote lung injury. Overexpression of FXYD5, a tissue-specific regulator of the Na,K-ATPase, in mice, impairs the
Sergey B Akopov et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 17(10), 1042-1049 (2006-10-05)
Identification of insulators is one of the most difficult problems in functional mapping of genomes. For this reason, up to now only a few insulators have been described. In this article we suggest an approach that allows direct isolation of
Yoshinori Ino et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(1), 365-370 (2002-01-05)
We report the cloning and characterization of a cancer-associated cell membrane glycoprotein recognized by mAb NCC-3G10. The antibody showed strong reactivity to a wide variety of cancer cells, but only to a limited number of normal cells including lymphocytes, endothelial
Kenji Ono et al.
Anticancer research, 30(9), 3273-3278 (2010-10-15)
Adjuvant chemotherapy is required following the resection of aggressive NSCLC. It is therefore necessary to identify factors that accurately predict prognosis. Tumor specimens were collected from 107 patients who underwent a complete resection for NSCLC from 1994-2000 in this Department.
Nieves María Gabrielli et al.
Fertility and sterility, 96(3), 554-561 (2011-07-22)
To study expression of dysadherin in human testis, epididymis, and spermatozoa. Prospective study. Basic research laboratory. Testis, epididymis, and testicular spermatozoa from patients under treatment and semen from volunteer donors. Reverse transcription-polymerase chain reaction, immunohistochemistry, immunocytochemistry, and Western immunoblotting. Dysadherin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service