Skip to Content
Merck
All Photos(5)

Key Documents

HPA006882

Sigma-Aldrich

Anti-VAMP8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EDB, Anti-Endobrevin, Anti-Vesicle-associated membrane protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MEEASEGGGNDRVRNLQSEVEGVKNIMTQNVERILARGENLEHLRNKTEDLEATSEHFKTTSQKVARKFWWKNVKMI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAMP8(8673)

Immunogen

Vesicle-associated membrane protein 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VAMP8 (Vesicle-associated membrane protein 8) is a platelet secretion granules belonging to the vesicle-associated membrane protein (VAMP) gene family. It is localized in the platelets. It forms a SNARE (N-ethylmaleimide-sensitive factor attachment protein, SNAP receptor) complex by attaching to the syntaxin, SNAP-25. It is essentialy involved in membrane fusion and trafficking, including platelet protein release and exocytosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70141

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiya Ohnishi et al.
Biological & pharmaceutical bulletin, 45(11), 1609-1615 (2022-11-04)
Autophagy is a highly conserved intracellular degrading system and its dysfunction is considered related to the cause of neurodegenerative disorders. A previous study showed that the inhibition of endocytosis transport attenuates soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) protein transport
János Polgár et al.
Blood, 100(3), 1081-1083 (2002-07-20)
Secretion of platelet granules is necessary for normal hemostasis. Platelet secretion requires soluble N-ethylmaleimide-sensitive factor attachment protein (SNAP) receptor (SNARE) complex formation between different members of the syntaxin, SNAP-25, and vesicle-associated membrane protein (VAMP) gene families. Using microcapillary reverse-phase high-performance
Joep F F Brinkman et al.
Molecular vision, 11, 582-586 (2005-08-20)
To select and characterize novel POAG disease genes. On the basis of genetic position (GLC1B), expression in the optic nerve, and biochemical function (targeted membrane transport processes), we selected the human VAMP5 and VAMP8 (encoding vesicle-associated membrane proteins 5 and
Hiroki Tanaka et al.
Journal of inherited metabolic disease, 45(6), 1191-1202 (2022-09-15)
Lysosomal storage disorders (LSDs) are inherited metabolic diseases caused by genetic defects in lysosomal enzymes or related factors. LSDs are associated with excessive accumulation of natural substrates in lysosomes leading to central nervous system and peripheral tissue damage. Abnormal autophagy
Virginie Hubert et al.
Biology open, 5(10), 1516-1529 (2016-09-16)
Autophagy is an evolutionarily conserved process used for removing surplus and damaged proteins and organelles from the cytoplasm. The unwanted material is incorporated into autophagosomes that eventually fuse with lysosomes, leading to the degradation of their cargo. The fusion event

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service