Skip to Content
Merck
All Photos(4)

Key Documents

HPA006421

Sigma-Aldrich

Anti-EMX1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Empty spiracles homolog 1 antibody produced in rabbit, Anti-Empty spiracles-like protein 1 antibody produced in rabbit, Anti-Homeobox protein EMX1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

RSLYGGPELVFPEAMNHPALTVHPAHQLGASPLQPPHSFFGAQHRDPLHFYPWVLRNRFFGHRFQASDVPQDGLLLHGPFARKPKRI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EMX1(2016)

General description

EMX1 (empty spiracles homeobox 1) gene is localized to human chromosome 2p14-p13. It is a late and narrow expression gene. It is a homeodomain transcription factor. It is a human ortholog of ems (empty spiracles) protein found in Drosophila. This protein is a resident of nucleus. In embryos, the expression of this gene is restricted to cerebral cortex and olfactory bulbs.

Immunogen

Homeobox protein EMX1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-EMX1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

EMX1 (empty spiracles homeobox 1) interacts with WDR11, which is a part of olfactory neuron development. In developing central nervous system, this protein determines cell fates. It participates in sonic hedgehog-Patched-Gli (Shh-Ptch-Gli) signaling pathway, where it acts as an effector protein for Gli-Kruppel family member 3. EMX1-lineage derived cells are responsible for two types of neuronal population in mature basal forebrain namely, a subclass of distinct amygdala excitatory neurons, and medium spiny neurons (MSNs) in the striatum. Thus, this protein determines the striatum and amygdala fate of developing neurons.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70921

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Cocas et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(50), 15933-15946 (2009-12-18)
In the developing mammalian basal telencephalon, neural progenitors from the subpallium generate the majority of inhibitory medium spiny neurons (MSNs) in the striatum, while both pallial- and subpallial-derived progenitors contribute to excitatory and inhibitory neuronal diversity in the amygdala. Using
K Kastury et al.
Genomics, 22(1), 41-45 (1994-07-01)
We have determined the chromosomal localization of four human homeobox-containing genes, EMX1, EMX2, OTX1, and OTX2, related to Drosophila genes expressed in the developing head of the fly. Murine homologs of these genes are expressed in specific nested domains in
A Simeone et al.
The EMBO journal, 11(7), 2541-2550 (1992-07-01)
We cloned two homeobox genes, Emx1 and Emx2, related to empty spiracles, a gene expressed in very anterior body regions during early Drosophila embryogenesis, and studied their expression in mouse embryos. Emx1 expression is detectable from day 9.5 of gestation
Hyung-Goo Kim et al.
American journal of human genetics, 87(4), 465-479 (2010-10-05)
By defining the chromosomal breakpoint of a balanced t(10;12) translocation from a subject with Kallmann syndrome and scanning genes in its vicinity in unrelated hypogonadal subjects, we have identified WDR11 as a gene involved in human puberty. We found six
Giorgia Quadrato et al.
Nature, 545(7652), 48-53 (2017-04-27)
In vitro models of the developing brain such as three-dimensional brain organoids offer an unprecedented opportunity to study aspects of human brain development and disease. However, the cells generated within organoids and the extent to which they recapitulate the regional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service