Skip to Content
Merck
All Photos(5)

Key Documents

HPA004726

Sigma-Aldrich

Anti-VCAN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CSPG2 antibody produced in rabbit, Anti-Chondroitin sulfate proteoglycan core protein 2, Anti-GHAP, Anti-Glial hyaluronate-binding protein, Anti-Large fibroblast proteoglycan, Anti-PG-M, Anti-Versican core protein precursor, AntiVCAN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SLSGKVSLPCHFSTMPTLPPSYNTSEFLRIKWSKIEVDKNGKDLKETTVLVAQNGNIKIGQDYKGRVSVPTHPEAVGDASLTVVKLLASDAGLYRCDVMYGIEDTQDTVSLTVDGVVFHYRAATSRYTLNFEAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCAN(1462)

General description

Versican (VCAN) encodes a large chondroitin sulfate proteoglycan named as versican. It is expressed in human fibroblasts. It consists of a hyaluronic acid-binding domain at the amino-terminal end. In the C termini, two epidermal growth factor (EGF) like repeats, a lectin-like sequence, and a complement regulatory protein-like domain are present. At the center, there is a glycosaminoglycan attachment site.

Immunogen

Versican core protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-VCAN antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Versican (VCAN) is involved in cell recognition. It connects extracellular matrix components and cell surface glycoproteins. It binds hyaluronan to connect proteins. As a result, it forms the large aggregate for maintaining structural integrity. Mutation in VCAN is responsible for Wagner syndrome (WS), an autosomal dominant disorder. It affects various ocular features.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86780

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yumiko Miyazaki et al.
Reproduction (Cambridge, England) (2018-11-06)
The endometrium extracellular matrix (ECM) is essential for embryo implantation. Versican, a large chondroitin sulfate proteoglycan that binds hyaluronan and forms large ECM aggregates, can influence fundamental physiological phenomena, such as cell proliferation, adhesion, and migration. The present study investigated
Svenja Hinderer et al.
Biomaterials, 35(7), 2130-2139 (2013-12-18)
Currently available heart valve replacements are limited in long-term performance or fail due to leaflet thickening, lack of growth or remodeling potential. In order to address these issues, it is necessary to mimic multiple factors of the native valvular extracellular
Chelsea Hope et al.
Journal of immunology (Baltimore, Md. : 1950), 199(5), 1933-1941 (2017-07-30)
Colorectal cancer originates within immunologically complex microenvironments. To date, the benefits of immunotherapy have been modest, except in neoantigen-laden mismatch repair-deficient tumors. Approaches to enhance tumor-infiltrating lymphocytes in the tumor bed may substantially augment clinical immunotherapy responses. In this article
Terence N Bukong et al.
Laboratory investigation; a journal of technical methods and pathology, 96(3), 361-374 (2016-01-12)
Little is known about the deposition and turnover of proteoglycans in liver fibrosis, despite their abundance in the extracellular matrix. Versican plays diverse roles in modulating cell behavior in other fibroproliferative diseases, but remains poorly described in the liver. Hepatic
Shun Chida et al.
Carcinogenesis, 37(9), 878-887 (2016-06-12)
To develop prognostic biomarkers that can discriminate stage II-III colorectal cancer patients with high risk of postoperative recurrence, we conducted a genome-wide screening of relapse-related genes utilizing multiple microarray cohorts. Among differentially expressed genes between tumor and nontumor, we identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service