Skip to Content
Merck

Skip To

F7387

Anti-Fibronectin Antibody

mouse monoclonal, FN-15

Synonym(s):

Anti-CIG, Anti-ED-B, Anti-FINC, Anti-FN, Anti-FNZ, Anti-GFND, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-SMDCF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
FN-15, monoclonal
Application:
ARR, ELISA (i), IF, WB
Citations:
72
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

FN-15, monoclonal

contains

15 mM sodium azide

species reactivity

mouse, human, chicken

technique(s)

indirect ELISA: 1:5,000, indirect immunofluorescence: 1:200 using cultured human fibroblasts, microarray: suitable, western blot: 1:800

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)

General description

Fibronectin (FN) is an extracellular matrix protein composed of two nearly-identical disulfide-bound polypeptides with typical molecular weights of 220-280 kDa. Cellular fibronectin is structurally and antigenically similar to cold insoluble globulin from plasma and antibodies to either form usually cross-react. Careful analysis of the FN molecule indicates that it contains several functionally and structurally distinct domains which may bind to cell surfaces and to a variety of molecules such as collagen, heparin, gelatin, fibrin and DNA. FN′s play an important role in diverse biological phenomena including cell adhesion, cell migration, hemostasis and thrombosis, wound healing and the ability to induce a more normal phenotype in transformed cells.
Monoclonal Anti-Fibronectin (mouse IgG1 isotype) is derived from the hybridoma produced the fusion of mouse myeloma cells and splenocytes from immunized mice. Fibronectin (FN) belongs to glycoprotein family widely expressed in many tissues. FN is an extracellular matrix protein composed of two nearly-identical disulfide-bound polypeptides with typical molecular weights of 220-240 kDa. There are three isoforms of fibronectin, that includes cellular FN, plasma FN and fetal FN. Cellular fibronectin is structurally and antigenically similar to cold insoluble globulin from plasma and antibodies to either form usually cross-react. Careful analysis of the FN molecule indicates that it contains several functionally and structurally distinct domains which may bind to cell surfaces and to a variety of molecules such as collagen, heparin, gelatin, fibrin and DNA. FN gene in human chromosome is mapped to 2q35.
Mouse monoclonal clone FN-15 anti-Fibronectin antibody labels fibronectin in the pericellular extracellular matrix of cultured human fibroblasts using indirect immunofluorescent methods. It also labels fibronectin in chicken and mouse cells. The FN-15 antibody specifically stains fibronectin in immunoblotting, and in an ELISA is reactive with human plasma fibronectin.

Immunogen

fibronectin from human plasma

Application

Monoclonal Anti-Fibronectin may be used to specifically localize fibronectin in human cell culture and in tissue sections by immunoblotting, immunohistochemical techniques and to stain fibronectin in hydrogels.
Mouse monoclonal clone FN-15 anti-Fibronectin antibody may be used to specifically localize fibronectin in human cell culture and in tissue sections by immunoblotting and immunohistochemical techniques. Monoclonal antibodies reacting specifically with FN may be used to localize FN in human cell cultures, in tissue sections and in plasma.

Biochem/physiol Actions

Fibronectin (FN) play an important role in diverse biological phenomena including cell adhesion, cell migration, hemostasis and thrombosis, wound healing and the ability to induce a more normal phenotype in transformed cells. FN is upregulated in many cancers. FN increases the expression of matrix metalloproteinases (MMPs), promoting cancer cell invasion and metastasis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0791F6140SAB4200784
species reactivity

mouse, human, chicken

species reactivity

goat, canine, human, chicken, sheep, rabbit, bovine

species reactivity

mouse, human, chicken

species reactivity

chicken, rat, human, mouse

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)

Gene Information

human ... FN1(2335)

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)

Gene Information

human ... FN1(2335)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

FN-15, monoclonal

clone

IST-3, monoclonal

clone

FN-3E2, monoclonal

clone

FN-3E2, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

culture supernatant


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Intracellular signaling cascades triggered by the NK1 fragment of hepatocyte growth factor in human prostate epithelial cell line PNT1A
Pavone LM, et al.
Cellular Signalling, 23(12), 1961-1971 (2011)
Jennifer Elisabeth Kloepper et al.
Experimental dermatology, 17(7), 592-609 (2008-06-19)
Since the discovery of epithelial hair follicle stem cells (eHFSCs) in the bulge of human hair follicles (HFs) an important quest has started: to define useful markers. In the current study, we contribute to this by critically evaluating corresponding published
Ehab El-Hallous et al.
The Journal of biological chemistry, 282(12), 8935-8946 (2007-01-27)
Fibrillin-containing microfibrils in elastic and nonelastic extracellular matrices play important structural and functional roles in various tissues, including blood vessels, lung, skin, and bone. Microfibrils are supramolecular aggregates of several protein and nonprotein components. Recently, a large region in the



Global Trade Item Number

SKUGTIN
F7387-.2ML04061833619605

Questions

Reviews

No rating value

Active Filters