Skip to Content
Merck
All Photos(6)

Key Documents

HPA002123

Sigma-Aldrich

Anti-ALDH1A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALDH-E1, Anti-ALHDII, Anti-Aldehyde dehydrogenase family 1 member A1, Anti-Aldehyde dehydrogenase, cytosolic, Anti-RALDH 1, Anti-RalDH1, Anti-Retinal dehydrogenase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

IYDEFVRRSVERAKKYILGNPLTPGVTQGPQIDKEQYDKILDLIESGKKEGAKLECGGGPWGNKGYFVQPTVFSNVTDEMRIAKEEIFGPVQQIMKFKSLDDVIKRANNTFYGL

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDH1A1(216)

Immunogen

Retinal dehydrogenase 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-ALDH1A1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Aldehyde dehydrogenase 1 family member A1 (ALDH1A1) belongs to the aldehyde dehydrogenase family. It plays a crucial role in ethanol metabolism in human. In addition to ethanol detoxification, it also has high impact on the metabolism of neurotransmitters and retinoic acid synthesis. It catalyzes the oxidation of endogenous and exogenous aldehydes depending on NAD(P)(+).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86228

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Yin et al.
Gastroenterology, 112(3), 766-775 (1997-03-01)
Alcohol dehydrogenase (ADH) and aldehyde dehydrogenase (ALDH) are the major enzymes responsible for ethanol metabolism in humans. The human stomach has been documented to be involved in the metabolism of first-passed alcohol. The aim of this study was to determine
Akos Z Szabo et al.
Stem cells (Dayton, Ohio), 31(4), 786-799 (2013-01-22)
Like for other somatic tissues, isolation of a pure population of stem cells has been a primary goal in epidermal biology. We isolated discrete populations of freshly obtained human neonatal keratinocytes (HNKs) using previously untested candidate stem cell markers aldehyde
N A Sophos et al.
Chemico-biological interactions, 130-132(1-3), 323-337 (2001-04-18)
Aldehyde dehydrogenase (ALDH) superfamily represents a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of endogenous and exogenous aldehydes. With the advent of megabase genome sequencing, the ALDH superfamily is expanding rapidly on many fronts. As
Eric Gyan et al.
Annals of diagnostic pathology, 51, 151696-151696 (2021-01-19)
Aldehyde dehydrogenase 1 member A1 (ALDH1A1) is one of the most well studied breast cancer stem cells. Its expression has been associated with poor clinicopathological features and clinical outcomes in several studies. This paper studies the expression of ALDH1A1 and
Riccardo Avvisati et al.
Cell reports, 43(4), 114080-114080 (2024-04-07)
Midbrain dopamine neurons are thought to play key roles in learning by conveying the difference between expected and actual outcomes. Recent evidence suggests diversity in dopamine signaling, yet it remains poorly understood how heterogeneous signals might be organized to facilitate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service