Skip to Content
Merck
All Photos(1)

Key Documents

H6508

Sigma-Aldrich

Heparin−Agarose

Type I, saline suspension

Synonym(s):

Heparin beads, Heparin resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

heparin from Porcine intestinal mucosa

Quality Level

type

Type I

form

saline suspension

extent of labeling

400-1500 μg per mL packed gel (activity approx. 140 USP units per mg)

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

suitability

suitable for chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Heparin-agarose is developed from porcine intestinal mucosa and is used in affinity chromatography. Heparin-agarose has been used in studies to provide information on human monocytic ehrlichiosis, tumor necrosis and the effects of coagulation from Vipera snake venom.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Silvestri et al.
Scandinavian journal of immunology, 53(3), 282-289 (2001-03-17)
An investigation was performed into the heparin-binding properties of a synthetic peptide deduced from the sequence of human cytomegalovirus glycoprotein B. The peptide, T7-13:3, amino acids 69-78, which was previously shown to contain a neutralization epitope was able to bind
D A Willins et al.
The Journal of biological chemistry, 266(17), 10768-10774 (1991-06-15)
Exogenous leucine affects the expression of a number of different operons in Escherichia coli. For at least some of these operons, the leucine-related effect is mediated by a protein called Lrp (Leucine-responsive regulatory protein). The purification of Lrp to near
I V Kaplan et al.
Atherosclerosis, 158(2), 455-463 (2001-10-05)
Oxidation of beta-lipoproteins has been linked to the development of arteriosclerosis. Using a copper mediated cell free system to oxidize beta-lipoproteins, we found that beta -lipoproteins isolated from plasma were less susceptible to oxidation than lipoproteins from serum and that
Analysis of Adeno-Associated Virus and HPV Interaction.
Hermonat, P.L., et al.
Methods in Molecular Medicine, 119, 397-409 (2006)
L C Andersen et al.
The Journal of experimental medicine, 173(1), 181-192 (1991-01-01)
Class II genes of the human major histocompatibility complex (MHC) are highly polymorphic. Allelic variation of structural genes provides diversity in immune cell interactions, contributing to the formation of the T cell repertoire and to susceptibility to certain autoimmune diseases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service