Skip to Content
Merck
All Photos(1)

Documents

H3667

Sigma-Aldrich

Human Serum

Heat Inactivated, from human male AB plasma, USA origin, sterile-filtered

Synonym(s):

HI Human Plasma, HI Human Sera, HI Human Serum, human plasma, human sera, human serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

human male plasma (Type AB)

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Serum is mostly required as a supplement to cell culture media. It contains macromolecules, carrier proteins, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors.

Application

Human Serum has been used as a supplement in culture media for monocytes. It has been used as a control in western blotting to study flightless I expression in serum.

Packaging

Packaging may vary for different batches due to short supply of the bottles used to pack this product. The presence or lack of shrink wrap around the cap of the bottle doesn’t impact product integrity/sterility of the product.

Analysis Note

Endotoxin tested

Other Notes

It is not unusual for our human serum products to be cloudy. This product is from pooled serum and cloudiness indicates that lipids are present in the serum. No clear, lipid-free human serum products are available.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A human and animal model-based approach to investigating the anti-inflammatory profile and potential of the 5-HT2B receptor antagonist AM1030.
Palmqvist N et al.
Journal of Inflammation (London, England), 13, 20-20 (2016)
Cytoskeletal protein Flightless I inhibits apoptosis, enhances tumor cell invasion and promotes cutaneous squamous cell carcinoma progression.
Kopecki Z et al.
Oncotarget, 6, 36426-36426 (2015)
Christina Megli et al.
The Journal of experimental medicine, 218(1) (2020-09-26)
The human placenta is a dynamic organ that modulates physiological adaptations to pregnancy. To define the immunological signature of the human placenta, we performed unbiased profiling of secreted immune factors from human chorionic villi isolated from placentas at mid and
Anneke H Hellinga et al.
International journal of molecular sciences, 21(23) (2020-12-03)
Bovine immunoglobulin G (bIgG) was previously shown to enhance innate immune responses to toll-like receptor (TLR) stimulation, via induction of trained immunity. In this study, we investigated whether minimally processed dairy streams with high levels of whey proteins as potential
Anubha Seth et al.
Cellular signalling, 68, 109538-109538 (2020-01-14)
Mammalian adenylate cyclases (ACs) are pseudoheterodimers with dissimilar hexahelical membrane-anchors, isoform-specifically conserved for more than half a billion years. We exchanged both membrane anchors of the AC isoform 2 by the quorum-sensing receptor from Vibrio harveyi, CqsS, which has a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service