Skip to Content
Merck
All Photos(1)

Key Documents

H5667

Sigma-Aldrich

Human Serum

Heat Inactivated (from male AB clotted whole blood), USA origin, sterile-filtered

Synonym(s):

HI Human sera, HI Human serum, HI OTC, Human Serum, human sera, off the clot, otc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

human

Quality Level

sterility

sterile-filtered

composition

Hemoglobin, ≤25 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

impurities

≤10 EU/mL Endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Human AB serum is used in tissue engineering, transplantation and cell therapy applications for the expansion of mesenchymal stem cells (MSC) from adipose tissue or mesenchymal stromal cells from human bone marrow; for standardized limbal epithelial stem cell graft generation and transplantation; for ex vivo expansion of NK cells from peripheral blood in Hematopoeitic Stem Cell Expansion Medium; and for upgrading pretransplant human islet culture technology.

Packaging

Packaging may vary for different batches due to short supply of the bottles used to pack this product. The presence or lack of shrink wrap around the cap of the bottle doesn’t impact product integrity/sterility of the product.

Warning

Each donor is tested for and found non-reactive for Hepatitis B & C and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Analysis Note

Endotoxin tested

Other Notes

It is not unusual for our human serum products to be cloudy. This product is from pooled serum and cloudiness indicates that lipids are present in the serum. No clear, lipid-free human serum products are available.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsvetana Petrova et al.
Scientific reports, 10(1), 3479-3479 (2020-02-28)
Type 2 Innate lymphoid cells (ILC2s) are implicated in helminth infections and asthma where they play a role in the production of Th2-type cytokines. ILC2s express the IL-33 receptor and are a major cell type thought to mediate the effects
Julie Kerr-Conte et al.
Transplantation, 89(9), 1154-1160 (2010-01-26)
BACKGROUND.: The original Edmonton protocol used fresh islets, but for obvious logistic advantages most transplant centers have implemented pretransplant culture in human albumin. The aim of this study was to improve current pretransplant human islet culture techniques. METHODS.: Clinical-grade purified
Torkild Visnes et al.
Nucleic acids research, 48(21), 12234-12251 (2020-11-20)
Altered oncogene expression in cancer cells causes loss of redox homeostasis resulting in oxidative DNA damage, e.g. 8-oxoguanine (8-oxoG), repaired by base excision repair (BER). PARP1 coordinates BER and relies on the upstream 8-oxoguanine-DNA glycosylase (OGG1) to recognise and excise
Nathan P Wiederhold et al.
Antimicrobial agents and chemotherapy, 52(11), 4145-4148 (2008-09-17)
Pyrosequencing was compared to Sanger dideoxy sequencing to detect mutations in FKS1 responsible for reduced echinocandin susceptibility in Candida albicans. These methods were in complete agreement for 10 of 12 clinical isolates with elevated echinocandin MICs, supporting the potential feasibility
Olívia Bacellar et al.
The Journal of infectious diseases, 200(1), 75-78 (2009-05-30)
Interleukin 17 (IL-17) plays a critical role in inflammation and autoimmunity. Very little is known about IL-17 in protozoa infection. Here, we show that lymphocytes obtained from patients with mucosal leishmaniasis and cutaneous leishmaniasis produce higher levels of IL-17 than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service