Skip to Content
Merck
All Photos(1)

Key Documents

H2414

Sigma-Aldrich

Anti-Humanin (HN) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Humanin Detection Antibody, Rabbit Anti-Humanin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:500

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MT-RNR2(4550)

General description

Humanin (HN) is a small polypeptide, derived from mitochondria. This protein is secreted by various cells and is localized in the plasma and bound to the cell membranes.

Immunogen

peptide corresponding to the humanin protein (amino acids 9-24).

Application

Anti-Humanin (HN) antibody produced in rabbit has been used in:
  • immunohistochemistry
  • immunoprecipitation (1:300)
  • fluorescent immunohistochemistry (1:300)
  • immunoblotting (1:500)

Biochem/physiol Actions

Humanin (HN) acts as a neuroprotective factor. It is involved in promoting cell survival. Humanin suppresses cell death in the neuronal cells caused by Alzheimer′s disease-related insults like anti-amyloid- precursor protein antibody, neurotoxic amyloid β (Aβ) peptides, and familial Alzheimer disease proteins.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changhan Lee et al.
Trends in endocrinology and metabolism: TEM, 24(5), 222-228 (2013-02-14)
Mitochondria have been largely considered as 'end-function' organelles, servicing the cell by producing energy and regulating cell death in response to complex signals. Being cellular entities with vital roles, mitochondria communicate back to the cell and actively engage in determining
Xingshun Xu et al.
Stroke, 37(10), 2613-2619 (2006-09-09)
Humanin (HN) is a 24-amino acid peptide best known for its ability to protect neurons from damage caused by Alzheimer disease-related proteins. This study examines the neuroprotective effects of HNG (a potent form of HN) on focal cerebral ischemia/reperfusion injury
Radhika H Muzumdar et al.
PloS one, 4(7), e6334-e6334 (2009-07-23)
Decline in insulin action is a metabolic feature of aging and is involved in the development of age-related diseases including Type 2 Diabetes Mellitus (T2DM) and Alzheimer's disease (AD). A novel mitochondria-associated peptide, Humanin (HN), has a neuroprotective role against
Mariela A Moreno Ayala et al.
Scientific reports, 10(1), 8542-8542 (2020-05-24)
Humanin (HN) is a mitochondrial-derived peptide with cytoprotective effect in many tissues. Administration of HN analogs has been proposed as therapeutic approach for degenerative diseases. Although HN has been shown to protect normal tissues from chemotherapy, its role in tumor
Yue Jia et al.
Biology of reproduction, 104(3), 717-730 (2020-12-18)
Humanin (HN) is a mitochondrial-derived peptide that protects many cells/tissues from damage. We previously demonstrated that HN reduces stress-induced male germ cell apoptosis in rodents. HN action in neuronal cells is mediated through its binding to a trimeric cell membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service