GERPN4201
AMDEX™ Sheep Anti Mouse IgG-HRP
Cytiva RPN4201
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
form
liquid
packaging
pkg of 0.5 mL
manufacturer/tradename
Cytiva RPN4201
storage temp.
2-8°C
General description
Sheep Anti-Mouse IgG-HRP. Sheep anti-mouse IgG conjugated to multiple HRP molecules on a Dextran backbone.
Features and Benefits
- It can be used in direct substitution for a standard anti-mouse HRP conjugate, without the need to alter existing protocols.
- It is important to optimize the dilution by setting up a range test.
- Option to reduce incubation times.
- Choice of six novel reagents.
- Well-controlled nonspecific binding properties.
Storage and Stability
Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
AMDEX is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service