Skip to Content
Merck
All Photos(1)

Key Documents

GERPN2235

ECL Select Western Blotting Detection Reagent

Cytiva RPN2235

Synonym(s):

chemiluminescent detection reagent, Western blotting detection reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva RPN2235

Related Categories

General description

Amersham ECL Select: Western blotting detection reagent with highly intense signal output, for detection of medium to very low protein levels.

Amersham ECL Select is a high sensitivity reagent for chemiluminescent Western blotting detection. High signal intensity makes Amersham ECL Select suitable for the most demanding Western blotting applications, allowing for detection of minute protein quantities. Its long lasting signals allow for multiple exposures and a convenient time window between experiment and analysis. Signals can be detected with CCD camera or X-ray film.

Amersham ECL Select typically exhibits a dynamic range that is complementary to Amersham ECL Prime, and similar to Amersham ECL Advance.

Application

Western Blotting

Features and Benefits

  • Improved sensitivity: Produces a very high signal intensity
  • Quantitative measurements: Linear dynamic range allowing for quantitation of medium to very low abundant proteins
  • Works with a wide range of antibody dilutions, reducing the need for lengthy optimization
  • Optimized for imaging with ImageQuant LAS 4000 (CCD-based imaging) and compatible with Amersham Hyperfilm
  • Compatible with Rainbow Molecular Weight Markers and Amersham ECL DualVue Western Blotting Markers.
  • Optimized for use with Amersham Hybond-P (PVDF) membranes and compatible with Amersham Hybond® ECL (nitrocellulose) membranes

Components

includes
Solution A (Luminol solution, 50 mL)
Solution B (Peroxide solution, 50 mL)
sufficient for 1000 cm2 membrane

complete packsize
for 1000 cm2 membrane

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Rainbow is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service