GERPN2109
ECL™ Blotting Reagents
Cytiva RPN2109
Synonym(s):
ECL detection reagents
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
wetted part: no
packaging
pkg of 1 kit
manufacturer/tradename
Cytiva RPN2109
color
Detection reagent 1: Colorless Detection reagent 2: Colorless
storage temp.
2-8°C
General description
Amersham™ ECL™ Western Blotting Detection Reagents were the world′s first commercial chemiluminescent reagents for Western blotting and are still the most widely used. They are at least 10-fold more sensitive than colorimetric assays and produce excellent and reliable results every time.
These reagents use enhanced luminol-based detection which is suitable for all routine western blotting. ECL™ Western Blotting Detection Reagents in combination with Hybond® ECL™ membranes form a complete solution for Western blot applications.
Contents include the following:
GERPN2109, 62.5 mL each of ECL™ Detection Reagents 1 and 2 (1000 cm2)
GERPN2209, 125 mL each (2000 cm2)
GERPN2106, 250 mL each (4000 cm2)
GERPN2134, 375 mL each (6000 cm2).
These reagents use enhanced luminol-based detection which is suitable for all routine western blotting. ECL™ Western Blotting Detection Reagents in combination with Hybond® ECL™ membranes form a complete solution for Western blot applications.
Contents include the following:
GERPN2109, 62.5 mL each of ECL™ Detection Reagents 1 and 2 (1000 cm2)
GERPN2209, 125 mL each (2000 cm2)
GERPN2106, 250 mL each (4000 cm2)
GERPN2134, 375 mL each (6000 cm2).
Emmission duration max. 2 h
Application
Western Blotting
Features and Benefits
- High sensitivity, ~10 pg in model system.
- Proven performance, the most frequently cited ECL™ Western Blotting Detection Reagents in publications
- Detectable on Hyperfilm ECL™ or with CCD imager
- Recommended membrane Hybond ECL™
Components
Sufficient for detection of 1000 cm2 of membrane:
62.5 mL Detection reagent 1(color : colorless)
62.5 mL Detection reagent 2 (color: colorless)
62.5 mL Detection reagent 1(color : colorless)
62.5 mL Detection reagent 2 (color: colorless)
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 12(7), e0180514-e0180514 (2017-07-12)
Mycoplasma infection has been reported in immunocompromised cancer patients; nevertheless, it is not clear if persistent Mycoplasma infection could facilitate the proliferation of cancer cells in immunocompromised organisms. The aim of this study was to examine the relationship between persistent
Journal of Cancer, 12(6), 1826-1837 (2021-02-23)
Decreased expression of proapoptotic genes can lead to the chemoresistenance in cancer therapy. Carboxyl-terminal binding protein 1 (CtBP1), a transcriptional corepressor with multiple oncogenic effects, has been previously identified to suppress the expression of two proapoptotic genes [BAX (BCL2 associated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service