GE29-0486-31
HisTrap™ Fast Flow Crude
Cytiva 29-0486-31, pack of 1 mL
About This Item
Recommended Products
packaging
pack of 1 mL
manufacturer/tradename
Cytiva 29-0486-31
availability
not available in North America
parameter
42 psi (H2O at room temperature.)
bed size
7 mm × 25 mm
bed volume
1 mL
column I.D.
7 mm
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning
2-14(Ni2+-stripped medium.)
working range
3-12(Ni2+-stripped medium.)
capacity
~40 mg binding capacity (histidine-tagged protein/ml medium)(Protein binding capacity is protein-to-protein dependent.)
~40 mg binding capacity(histidine-tagged protein)
suitability
suitable for bioprocess medium
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Optimized for purification of histidine-tagged proteins directly from homogenized, unclarified cell lysates.
- High binding capacity, approx. 40 mg/ml medium and negligible Ni2+ leakage
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives.
- Reduced sample preparation time, which minimizes risk of protein degradation by proteases.
- No need for centrifugation or filtration of the samples. Direct sample application.
- Reliable purification of histidine-tagged proteins directly from unclarified lysates–simply sonicate and run.
- Reduced sample preparation time, which minimizes risk of protein degradation by proteases
- Simple operation with a syringe, pump, or chromatography system such as ÄKTAdesign or FPLC System
- Permit rapid yet reliable separations with a minimum of sample preparation and equipment
Storage and Stability
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service