Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9537-66

HiTrap® TALON® Crude

Cytiva 28-9537-66, pack of 5 × 1 mL

Synonym(s):

Crude TALON Column, HiTrap TALON, TALON Crude Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

TALON® Superflow Cobalt-IMAC

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 28-9537-66

parameter

1 ml/min - 4 ml/min flow rate
3 bar

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

60-160 μm

cleaning

2-14(Co2+-stripped medium)

working range

3-12(Co2+-stripped medium)

capacity

≥20 mg binding capacity (histidine-tagged protein)

storage temp.

2-8°C

Related Categories

General description

HiTrap® Talon® crude 1 mL and 5 mL columns allow for rapid separations and easy scale-up with a minimum of sample preparation and equipment.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Application

Talon® Superflow is a cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. Talon® Superflow and prepacked HiTrap® Talon® crude columns allow Protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.

HiTrap® Talon® crude columns allow for rapid separations and easy scale-up. Samples can be applied directly to the column after thorough cell disruption - without precentrifugation and filtration. The filters in the top and bottom of the column make it possible to directly load unclarified lysates on the column without causing back pressure problems or leakage of the Talon® Superflow beads. Prior to loading samples on HiTrap® Talon® crude columns, cell lysate should be prepared using methods such as sonication, homogenization, or freeze/thaw.

Features and Benefits

  • Columns prepacked with cobalt-based chromatography media designed for purification of histidine-tagged recombinant Proteins by immobilized metal affinity chromatography (IMAC).
  • Suitable for IMAC purification of histidine-tagged Proteins when Ni2+ is not the optimal choice of metal ion.
  • Compatible with commonly used IMAC reagents and appropriate for purifying Proteins under native or denaturing conditions.
  • Prepacked HiTrap® Talon® crude columns save time by allowing the application of unclarified samples directly, minimizing labor and degradation of Proteins.
  • HiTrap® Talon® crude columns allow repeated runs without stripping, cleaning, or Co2+ recharging.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Talon is a registered trademark of Clontech

Pictograms

FlameFlame

Signal Word

Warning-Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F

Flash Point(C)

38 - 43 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service