GE28-9165-37
HiTrap® Capto™ DEAE
Cytiva 28-9165-37, pack of 5 × 1 mL
About This Item
Recommended Products
ligand
diethylaminoethyl
description
Ion Exchanger Type (value)
packaging
pack of 5 × 1 mL
manufacturer/tradename
Cytiva 28-9165-37
parameter
<4 mL/min flow rate
42 psi
bed size
7 mm × 25 mm
bed volume
1 mL
column I.D.
7 mm
matrix
highly cross-linked agarose with dextran surface extender
average diameter
90 μm (d50v)
cleaning
2-14
working range
2-12
suitability
suitable for bioprocess medium
Looking for similar products? Visit Product Comparison Guide
Storage and Stability
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service