GE17-0921-07
IMAC Sepharose™ 6 Fast Flow
Cytiva 17-0921-07, pack of 25 mL
About This Item
Recommended Products
packaging
pack of 25 mL
manufacturer/tradename
Cytiva 17-0921-07
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning in place
2-14(Medium uncharged with metal ions.)
working range
3-12
capacity
~40 mg binding capacity (histidine-tagged protein/ml medium when changed with Ni2+)(Protein- and metal-ion dependent.)
suitability
suitable for bioprocess medium
Related Categories
General description
Application
IMAC Sepharose™ 6 Fast FLow is composed of cross-linked 6% Agarose beads modified with a novel chelating ligand immobilized to the base matrix. IMAC Sepharose™ 6 Fast FLow is supplied free of metal ions, allowing the user to charge it with the most appropriate metal ion for purification of a target Protein.
The coupling chemistry and the novel ligand is designed to give minimal ligand leakage while providing high capacity.
IMAC Sepharose™ 6 Fast FLow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
As member of the BioProcess media range, IMAC Sepharose™ 6 Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support
Features and Benefits
- Uncharged IMAC resin allowing possibility to charge with your metal ion of choice for optimized selectivity
- Convenient purification of histidine-tagged Proteins, also when Ni2+ is not the best choice of metal ion
- As BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
- Available in the convenient and time-saving prepacked HiTrap® and HiPrep™ formats.
Storage and Stability
Analysis Note
Legal Information
These products are sold under a license from Sigma-Aldrich under patent number EP 1276716 (Metal chelating compositions) and equivalent patents and patent applications in other countries.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service